Search
Search

LpR1| Gene Symbol: LpR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104461.3 | NP_001097931.3 | 10 - 11 | 2877 | 100 | |
| NM_001104462.1 | NP_001097932.1 | 9 - 10 | 2706 | 100 | ||
| NM_001104463.3 | NP_001097933.2 | 10 - 11 | 3189 | 100 | ||
| NM_001104464.2 | NP_001097934.2 | 12 - 13 | 3429 | 100 | ||
| NM_001170263.2 | NP_001163734.1 | 10 - 11 | 3135 | 100 | ||
| NM_001276043.1 | NP_001262972.1 | 10 - 11 | 3129 | 100 | ||
| NM_001276044.1 | NP_001262973.1 | 11 - 12 | 5349 | 100 | ||
| NM_001276045.1 | NP_001262974.1 | 9 - 10 | 2623 | 100 | ||
| NM_170240.3 | NP_733119.2 | 10 - 11 | 2883 | 100 | ||
| NM_206573.2 | NP_996296.2 | 11 - 12 | 3312 | 100 | ||
| GenBank mRNA | BT011057.1 | - | 10 - 11 | 3189 | 100 | |
| BT011063.1 | - | 11 - 12 | 3291 | 100 | ||
| BT099946.1 | - | 9 - 10 | 2706 | 100 | ||
Set Membership: |
|