Search
Search

CanA1| Gene Symbol: CanA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260448.2 | NP_001247377.1 | 6 - 7 | 1293 | 58 | |
| NM_001260449.2 | NP_001247378.2 | 5 - 6 | 1749 | 58 | ||
| NM_079861.4 | NP_524600.3 | 6 - 7 | 1313 | 58 | ||
| GenBank mRNA | AY128480.1 | - | 6 - 7 | 1311 | 58 | |
Set Membership: |
|