Search
Search

lds| Gene Symbol: lds | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080111.4 | NP_524850.2 | 3 - 4 | 1665 | 69 | |
| GenBank mRNA | BT150462.1 | - | 3 - 4 | 1663 | 69 | |
| BT150466.1 | - | 3 - 4 | 1663 | 69 | ||
Set Membership: |
|