Search
Search

ftr67| Gene Symbol: ftr67 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039719.2 | NP_001034808.2 | 5 - 6 | 1205 | 99 | |
| GenBank mRNA | BC076205.1 | - | 5 - 6 | 1159 | 99 | |
| BC093187.1 | - | 5 - 6 | 1161 | 99 | ||
| BC114247.1 | - | 5 - 6 | 1190 | 99 | ||
Set Membership: |
|