Search
Search

ptgs2a| Gene Symbol: ptgs2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153657.1 | NP_705943.1 | 3 - 4 | 389 | 64 | |
| GenBank mRNA | AY028585.1 | - | 3 - 4 | 389 | 64 | |
| BC056736.1 | - | 3 - 4 | 378 | 64 | ||
| BC063232.1 | - | 3 - 4 | 364 | 64 | ||
Set Membership: |
|