Search
Search

ptpn4b| Gene Symbol: ptpn4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044313.2 | NP_001037778.2 | 4 - 5 | 364 | 75 | |
| XM_005173946.4 | XP_005174003.1 | 5 - 6 | 760 | 75 | ||
| XM_009296096.3 | XP_009294371.1 | 5 - 6 | 760 | 75 | ||
| GenBank mRNA | AW174241.1 | - | 4 - 5 | 356 | 75 | |
Set Membership: |
|