Search
Search

antxr2b| Gene Symbol: antxr2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102568.1 | NP_001096038.1 | 10 - 11 | 844 | 150 | |
| GenBank mRNA | EF591979.1 | - | 10 - 11 | 844 | 150 | |
Set Membership: |
|