Search
Search

ca5a| Gene Symbol: ca5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111201.1 | NP_001104671.1 | 3 - 4 | 865 | 64 | |
| XM_005163076.4 | XP_005163133.1 | 3 - 4 | 698 | 64 | ||
| GenBank mRNA | BC153994.1 | - | 3 - 4 | 865 | 64 | |
Set Membership: |
|