Search
Search

ALOX5AP| Gene Symbol: ALOX5AP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163965.1 | NP_001157437.1 | 2 - 3 | 229 | 57 | |
| XM_005601074.3 | XP_005601131.1 | 303 | 57 | |||
| XM_005601075.3 | XP_005601132.1 | 260 | 57 | |||
| XM_023621300.1 | XP_023477068.1 | 306 | 57 | |||
| XM_023621301.1 | XP_023477069.1 | 276 | 57 | |||
| XM_023621302.1 | XP_023477070.1 | 271 | 57 | |||
| GenBank mRNA | M96552.1 | - | 2 - 3 | 171 | 57 | |
Set Membership: |
|