Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ?
Create Account
Applications
Real-Time PCR
Oligos, Primers, Probes and Genes
Cloning
Protein Biology
Cell Culture and Transfection
Cell and Gene Therapy
Cell Analysis
Mass Spectrometry
Chromatography
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Online Order
Custom Primers & TaqMan Probes
miRNA Mimics & Inhibitors
Stealth RNAi / siRNA
Silencer Select siRNAs
Custom DNA Oligos
GeneArt Services
GeneArt Strings DNA Fragments
TrueGuide CRISPR gRNA
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all online orderable products
Services
Custom Services
Instrument Qualification Services
Technical Services
Pipette Services
Sample Request
Instrument Maintenance Services
Repairs and Relocation Services
OEM and Licensing Services
Events, Seminars, Training
Unity Lab Services
See all services
Support
Catalogs
Application Notes
Safety Data Sheets
Legal and Regulatory
Certificates of Analysis Search
Nunc/Nalgene Product Certificates
e-learning
FAQs
Learning Centers
Technical Support Centers
See all help and support topics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign In
Sign In
Don't have an account ?
Create Account
Connect Your Lab
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Ec06958710_g1
See other TTN GE Assays ›
Gene Symbol:
TTN
Gene Name:
titin
Gene Aliases:
-
Chromosome Location:
Chr.18: 56785112 - 57053971 on Build EquCab3.0
Species:
Horse
Species Specific ID (Flybase ID):
-
Assay ID
Ec06958710_g1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2,900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2,900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2,900 rxns
Availability
Made To Order
Catalog #
4351372
Price
4,430.00
Your Price
Online offer:
4430.0
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: TTN
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
XM_023622070.1
XP_023477838.1
88748
109
XM_023622071.1
XP_023477839.1
88736
109
XM_023622073.1
XP_023477841.1
88673
109
XM_023622074.1
XP_023477842.1
88670
109
XM_023622075.1
XP_023477843.1
88664
109
XM_023622076.1
XP_023477844.1
88664
109
XM_023622077.1
XP_023477845.1
88664
109
XM_023622078.1
XP_023477846.1
88664
109
XM_023622079.1
XP_023477847.1
88664
109
XM_023622080.1
XP_023477848.1
88664
109
XM_023622081.1
XP_023477849.1
88664
109
XM_023622082.1
XP_023477850.1
88646
109
XM_023622083.1
XP_023477851.1
88610
109
XM_023622084.1
XP_023477852.1
88592
109
XM_023622085.1
XP_023477853.1
88592
109
XM_023622086.1
XP_023477854.1
88589
109
XM_023622087.1
XP_023477855.1
88580
109
XM_023622088.1
XP_023477856.1
88580
109
XM_023622089.1
XP_023477857.1
88580
109
XM_023622090.1
XP_023477858.1
88577
109
XM_023622091.1
XP_023477859.1
88544
109
XM_023622092.1
XP_023477860.1
88496
109
XM_023622093.1
XP_023477861.1
88496
109
XM_023622094.1
XP_023477862.1
88496
109
XM_023622095.1
XP_023477863.1
88496
109
XM_023622096.1
XP_023477864.1
88493
109
XM_023622097.1
XP_023477865.1
88490
109
XM_023622098.1
XP_023477866.1
88490
109
XM_023622099.1
XP_023477867.1
88472
109
XM_023622100.1
XP_023477868.1
88460
109
XM_023622101.1
XP_023477869.1
88448
109
XM_023622102.1
XP_023477870.1
88748
109
XM_023622103.1
XP_023477871.1
88412
109
XM_023622104.1
XP_023477872.1
88409
109
XM_023622105.1
XP_023477873.1
88406
109
XM_023622106.1
XP_023477874.1
88406
109
XM_023622107.1
XP_023477875.1
88328
109
XM_023622108.1
XP_023477876.1
88325
109
XM_023622109.1
XP_023477877.1
88259
109
XM_023622110.1
XP_023477878.1
88244
109
XM_023622111.1
XP_023477879.1
88241
109
XM_023622112.1
XP_023477880.1
88238
109
XM_023622113.1
XP_023477881.1
88181
109
XM_023622114.1
XP_023477882.1
88160
109
XM_023622115.1
XP_023477883.1
88154
109
XM_023622116.1
XP_023477884.1
88085
109
XM_023622117.1
XP_023477885.1
88076
109
XM_023622118.1
XP_023477886.1
88007
109
XM_023622119.1
XP_023477887.1
87989
109
XM_023622120.1
XP_023477888.1
89345
109
Back To Top
More Information
Set Membership:
Amplicon length greater than or equal to 101
Equus caballus
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline