Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: TENM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204862.1 | NP_990193.1 | 23 - 24 | 4386 | 74 | |
XM_015278189.2 | XP_015133675.1 | 24 - 25 | 5648 | 74 | ||
XM_015278190.2 | XP_015133676.1 | 24 - 25 | 5645 | 74 | ||
XM_015278191.2 | XP_015133677.1 | 23 - 24 | 5624 | 74 | ||
GenBank mRNA | AJ238613.1 | - | 23 - 24 | 4386 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |