Search
Search

FCRL4| Gene Symbol: FCRL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097529.2 | NP_001090998.2 | 3 - 4 | 349 | 90 | |
| XM_015298625.2 | XP_015154111.2 | 5 - 6 | 2932 | 90 | ||
| XM_015298627.2 | XP_015154113.2 | 5 - 6 | 2919 | 90 | ||
| XM_015298629.2 | XP_015154115.2 | 5 - 6 | 2820 | 90 | ||
| XM_015298630.2 | XP_015154116.2 | 3 - 4 | 2760 | 90 | ||
| XM_015298631.2 | XP_015154117.2 | 4 - 5 | 3014 | 90 | ||
| XM_015298633.2 | XP_015154119.2 | 5 - 6 | 2541 | 90 | ||
| XM_015298634.2 | XP_015154120.2 | 6 - 7 | 2109 | 90 | ||
| XM_015298635.2 | XP_015154121.2 | 4 - 5 | 2146 | 90 | ||
| XM_015298636.2 | XP_015154122.2 | 5 - 6 | 2009 | 90 | ||
| XM_015298638.2 | XP_015154124.2 | 5 - 6 | 2527 | 90 | ||
| XM_025143401.1 | XP_024999169.1 | 4 - 5 | 859 | 90 | ||
| XM_025143402.1 | XP_024999170.1 | 5 - 6 | 751 | 90 | ||
| GenBank mRNA | AM412311.1 | - | 3 - 4 | 349 | 90 | |
Set Membership: |
|