Search
Search

THY1| Gene Symbol: THY1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204381.2 | NP_989712.2 | 3 - 4 | 484 | 87 | |
| GenBank mRNA | AY230132.1 | - | 2 - 3 | 378 | 87 | |
| CN224301.1 | - | 3 - 4 | 484 | 87 | ||
| CO420629.1 | - | 1 - 2 | 173 | 87 | ||
| JQ088097.1 | - | 1 - 2 | 180 | 87 | ||
| L14924.1 | - | 2 - 3 | 384 | 87 | ||
| S47368.1 | - | 2 - 3 | 384 | 87 | ||
Set Membership: |
|