Search
Search

TRAPPC2| Gene Symbol: TRAPPC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006263.1 | NP_001006263.1 | 3 - 4 | 278 | 128 | |
| XM_015273553.2 | XP_015129039.1 | 3 - 4 | 351 | 128 | ||
| GenBank mRNA | AJ720040.1 | - | 3 - 4 | 278 | 128 | |
Set Membership: |
|