Search
Search

RNASE6| Gene Symbol: RNASE6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205259.2 | NP_990590.1 | 2 | 742 | 163 | |
| XM_025151489.1 | XP_025007257.1 | 2 | 689 | 163 | ||
| GenBank mRNA | BX935629.1 | - | 2 | 696 | 163 | |
| DQ395275.1 | - | 2 | 742 | 163 | ||
| X61193.1 | - | 705 | 162 | |||
Set Membership: |
|