Search
Search

PK3
LOC100807615| Gene Symbol: PK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001249502.2 | NP_001236431.1 | 2 - 3 | 694 | 89 | |
| GenBank mRNA | AJ879897.1 | - | 2 - 3 | 696 | 89 | |
| AK244589.1 | - | 2 - 3 | 728 | 89 | ||
| Gene Symbol: LOC100807615 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK245940.1 | - | 2 - 3 | 827 | 89 | |
Set Membership: |
|