| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10021203.hg.1 |
ENST00000369887.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10021203.hg.1 |
ENST00000489268.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10021203.hg.1 |
NM_000102.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10021203.hg.1 |
TC10001625.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10021204.hg.1 |
ENST00000369887.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10021204.hg.1 |
NM_000102.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10021204.hg.1 |
TC10001625.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10021200.hg.1 |
ENST00000369887.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10021200.hg.1 |
ENST00000489268.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10021200.hg.1 |
NM_000102.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10021200.hg.1 |
TC10001625.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011696.hg.1 |
ENST00000369887.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011696.hg.1 |
ENST00000369887.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011696.hg.1 |
ENST00000369887.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011696.hg.1 |
ENST00000369887.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011696.hg.1 |
ENST00000369887.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011696.hg.1 |
ENST00000369887.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011696.hg.1 |
ENST00000369887.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011696.hg.1 |
ENST00000369887.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011696.hg.1 |
ENST00000369887.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011696.hg.1 |
ENST00000369887.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011696.hg.1 |
ENST00000369887.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011696.hg.1 |
ENST00000369887.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011696.hg.1 |
ENST00000469683.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011696.hg.1 |
ENST00000469683.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011696.hg.1 |
ENST00000469683.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011696.hg.1 |
ENST00000489268.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011696.hg.1 |
ENST00000489268.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011696.hg.1 |
ENST00000489268.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011696.hg.1 |
ENST00000489268.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011696.hg.1 |
ENST00000489268.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011696.hg.1 |
ENST00000489268.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011696.hg.1 |
ENST00000489268.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011696.hg.1 |
ENST00000489268.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011696.hg.1 |
NM_000102.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011696.hg.1 |
NM_000102.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011696.hg.1 |
NM_000102.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011696.hg.1 |
NM_000102.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011696.hg.1 |
NM_000102.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011696.hg.1 |
NM_000102.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011696.hg.1 |
NM_000102.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011696.hg.1 |
NM_000102.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011696.hg.1 |
NM_000102.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011696.hg.1 |
NM_000102.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011696.hg.1 |
NM_000102.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011696.hg.1 |
NM_000102.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10021202.hg.1 |
ENST00000369887.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10021202.hg.1 |
ENST00000489268.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10021202.hg.1 |
NM_000102.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10021202.hg.1 |
TC10001625.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC1000011696.hg.1 |
ENST00000369887.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011696.hg.1 |
ENST00000369887.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011696.hg.1 |
ENST00000369887.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011696.hg.1 |
ENST00000369887.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011696.hg.1 |
ENST00000369887.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011696.hg.1 |
ENST00000369887.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011696.hg.1 |
ENST00000369887.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011696.hg.1 |
ENST00000369887.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011696.hg.1 |
ENST00000369887.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011696.hg.1 |
ENST00000369887.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011696.hg.1 |
ENST00000369887.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011696.hg.1 |
ENST00000369887.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011696.hg.1 |
ENST00000469683.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011696.hg.1 |
ENST00000469683.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011696.hg.1 |
ENST00000469683.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011696.hg.1 |
ENST00000489268.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011696.hg.1 |
ENST00000489268.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011696.hg.1 |
ENST00000489268.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011696.hg.1 |
ENST00000489268.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011696.hg.1 |
ENST00000489268.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011696.hg.1 |
ENST00000489268.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011696.hg.1 |
ENST00000489268.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011696.hg.1 |
ENST00000489268.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011696.hg.1 |
NM_000102.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011696.hg.1 |
NM_000102.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011696.hg.1 |
NM_000102.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011696.hg.1 |
NM_000102.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011696.hg.1 |
NM_000102.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011696.hg.1 |
NM_000102.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011696.hg.1 |
NM_000102.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011696.hg.1 |
NM_000102.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011696.hg.1 |
NM_000102.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011696.hg.1 |
NM_000102.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011696.hg.1 |
NM_000102.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011696.hg.1 |
NM_000102.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000180069.hg.1 |
ENST00000369887.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000180069.hg.1 |
ENST00000489268.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000180069.hg.1 |
NM_000102.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000180069.hg.1 |
ENST00000369887.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000180069.hg.1 |
ENST00000489268.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000180069.hg.1 |
NM_000102.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000180069.hg.1 |
TC1000011696.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1000180065.hg.1 |
ENST00000369887.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000180065.hg.1 |
ENST00000489268.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000180065.hg.1 |
NM_000102.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000180065.hg.1 |
ENST00000369887.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000180065.hg.1 |
ENST00000489268.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000180065.hg.1 |
NM_000102.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000180065.hg.1 |
TC1000011696.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1000180070.hg.1 |
ENST00000369887.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000180070.hg.1 |
NM_000102.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000180070.hg.1 |
ENST00000369887.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000180070.hg.1 |
NM_000102.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000180070.hg.1 |
TC1000011696.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1000180067.hg.1 |
ENST00000369887.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000180067.hg.1 |
ENST00000489268.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000180067.hg.1 |
NM_000102.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000180067.hg.1 |
ENST00000369887.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000180067.hg.1 |
ENST00000489268.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000180067.hg.1 |
NM_000102.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000180067.hg.1 |
TC1000011696.hg.1 |
Clariom D TC |