Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00295149_s1
          See other CDC42EP3 GE Assays ›
          Gene Symbol:
          CDC42EP3
          Gene Name:
          CDC42 effector protein 3
          Gene Aliases:
          BORG2, CEP3, UB1
          Chromosome Location:
          Chr.2: 37641882 - 37672535 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00295149_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price 4,565.00
          Your Price
          Online offer:
          4565.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CDC42EP3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001270436.1 NP_001257365.1 2 3721 63
          NM_001270437.1 NP_001257366.1 2 3677 63
          NM_001270438.1 NP_001257367.1 2 3653 63
          NM_006449.4 NP_006440.2 2 4312 63
          XM_011532482.2 XP_011530784.1 2 4118 63
          GenBank mRNA AK055915.1 - 1 851 63
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BU194600.1 - 1 206 Chr. 2 - 37642669 - 37643479 [-] on Build GRCh38 63
          GenBankmRNA R19529.1 - 1 Chr. 2 - 37643113 - 37643429 [-] on Build GRCh38 63
          GenBankmRNA DA252943.1 - 1 404 Chr. 2 - 37643117 - 37643687 [-] on Build GRCh38 63
          GenBankmRNA CD242069.1 - 1 128 Chr. 2 - 37642744 - 37643412 [-] on Build GRCh38 63
          GenBankmRNA CN417889.1 - 1 552 Chr. 2 - 37643101 - 37643837 [-] on Build GRCh38 63
          GenBankmRNA R13631.1 - 90 Chr. - - [-] on Build 63
          GenBankmRNA AL133837.1 - 1 672 Chr. 2 - 37643242 - 37643957 [-] on Build GRCh38 63
          GenBankmRNA BG168552.1 - 1 490 Chr. 2 - 37643169 - 37643766 [-] on Build GRCh38 63
          GenBankmRNA CF121340.1 - 1 347 Chr. 2 - 37642926 - 37643623 [-] on Build GRCh38 63

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream glutaminyl-peptide cyclotransferase QPCT GCT,
          QC,
          sQC
          -268.6kb Anti-sense Chr. 2 - 37344610 - 37373322 on NCBI Build GRCh38
          Downstream long intergenic non-protein coding RNA 211 LINC00211 NCRNA00211 182.1kb Sense Chr. 2 - 37826247 - 37875863 on NCBI Build GRCh38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene CDC42EP3

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          Rho protein signal transduction
          regulation of cell shape
          positive regulation of actin filament polymerization
          positive regulation of pseudopodium assembly
          positive regulation of GTPase activity
          GTPase activator activity
          cytoskeletal regulatory protein binding
          GTP-Rho binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0200007265.hg.1 ENST00000419425.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007265.hg.1 ENST00000419425.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007265.hg.1 ENST00000419425.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007265.hg.1 ENST00000419425.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200007265.hg.1 ENST00000419425.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200007265.hg.1 ENST00000419425.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200007265.hg.1 ENST00000419425.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200007265.hg.1 ENST00000419425.1 Ensembl
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 104054 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline