| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016157.hg.1 |
ENST00000368012.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016157.hg.1 |
ENST00000368012.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016157.hg.1 |
ENST00000368012.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016157.hg.1 |
ENST00000368012.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016157.hg.1 |
ENST00000368012.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016157.hg.1 |
ENST00000368012.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016157.hg.1 |
ENST00000368012.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016157.hg.1 |
ENST00000368012.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016157.hg.1 |
ENST00000368012.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016157.hg.1 |
ENST00000368012.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016157.hg.1 |
ENST00000368012.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016157.hg.1 |
ENST00000368012.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016157.hg.1 |
ENST00000368012.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016157.hg.1 |
ENST00000368012.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016157.hg.1 |
ENST00000368012.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016157.hg.1 |
ENST00000486694.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016157.hg.1 |
ENST00000486694.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016157.hg.1 |
ENST00000486694.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016157.hg.1 |
ENST00000486694.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016157.hg.1 |
NM_030916.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016157.hg.1 |
NM_030916.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016157.hg.1 |
NM_030916.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016157.hg.1 |
NM_030916.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016157.hg.1 |
NM_030916.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016157.hg.1 |
NM_030916.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016157.hg.1 |
NM_030916.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016157.hg.1 |
NM_030916.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016157.hg.1 |
NM_030916.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016157.hg.1 |
NM_030916.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016157.hg.1 |
NM_030916.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016157.hg.1 |
NM_030916.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016157.hg.1 |
NM_030916.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016157.hg.1 |
NM_030916.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016157.hg.1 |
NM_030916.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01053477.hg.1 |
ENST00000368012.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01053477.hg.1 |
ENST00000453926.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01053477.hg.1 |
NM_030916.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01053477.hg.1 |
TC01003415.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0100016157.hg.1 |
ENST00000368012.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016157.hg.1 |
ENST00000368012.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016157.hg.1 |
ENST00000368012.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016157.hg.1 |
ENST00000368012.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016157.hg.1 |
ENST00000368012.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016157.hg.1 |
ENST00000368012.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016157.hg.1 |
ENST00000368012.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016157.hg.1 |
ENST00000368012.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016157.hg.1 |
ENST00000368012.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016157.hg.1 |
ENST00000368012.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016157.hg.1 |
ENST00000368012.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016157.hg.1 |
ENST00000368012.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016157.hg.1 |
ENST00000368012.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016157.hg.1 |
ENST00000368012.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016157.hg.1 |
ENST00000368012.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016157.hg.1 |
ENST00000486694.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016157.hg.1 |
ENST00000486694.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016157.hg.1 |
ENST00000486694.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016157.hg.1 |
ENST00000486694.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016157.hg.1 |
NM_030916.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016157.hg.1 |
NM_030916.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016157.hg.1 |
NM_030916.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016157.hg.1 |
NM_030916.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016157.hg.1 |
NM_030916.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016157.hg.1 |
NM_030916.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016157.hg.1 |
NM_030916.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016157.hg.1 |
NM_030916.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016157.hg.1 |
NM_030916.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016157.hg.1 |
NM_030916.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016157.hg.1 |
NM_030916.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016157.hg.1 |
NM_030916.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016157.hg.1 |
NM_030916.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016157.hg.1 |
NM_030916.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016157.hg.1 |
NM_030916.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100222967.hg.1 |
ENST00000368012.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100222967.hg.1 |
NM_030916.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100222967.hg.1 |
ENST00000368012.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100222967.hg.1 |
NM_030916.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100222967.hg.1 |
TC0100016157.hg.1 |
Clariom D TC |