Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00418886_m1
          See other LINC00644 GE Assays ›
          Gene Symbol:
          LINC00644
          Gene Name:
          long intergenic non-protein coding RNA 644
          Gene Aliases:
          -
          Chromosome Location:
          Chr.14: 62134149 - 62139973 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00418886_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LINC00644
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_104063.1 1 - 2 64 91

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1400009375.hg.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Recommended Assay Homo sapiens Inventoried Probe spans exons

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1400009375.hg.1 ENST00000555156.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400009375.hg.1 ENST00000555156.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400009375.hg.1 NR_104063.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400009375.hg.1 NR_104063.1 Refseq
          Affymetrix Human Clariom D Assay TC1400009375.hg.1 ENST00000555156.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400009375.hg.1 ENST00000555156.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400009375.hg.1 NR_104063.1 Refseq
          Affymetrix Human Clariom D Assay TC1400009375.hg.1 NR_104063.1 Refseq
          Affymetrix Human Clariom D Assay PSR1400169278.hg.1 ENST00000555156.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1400169278.hg.1 NR_104063.1 Refseq
          Affymetrix Human Clariom D Assay PSR1400169278.hg.1 ENST00000555156.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1400169278.hg.1 NR_104063.1 Refseq
          Affymetrix Human Clariom D Assay PSR1400169278.hg.1 TC1400009375.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1400169279.hg.1 ENST00000555156.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1400169279.hg.1 NR_104063.1 Refseq
          Affymetrix Human Clariom D Assay PSR1400169279.hg.1 ENST00000555156.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1400169279.hg.1 NR_104063.1 Refseq
          Affymetrix Human Clariom D Assay PSR1400169279.hg.1 TC1400009375.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline