Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00735290_m1
          See other VSIR GE Assays ›
          Gene Symbol:
          VSIR
          Gene Name:
          V-set immunoregulatory receptor
          Gene Aliases:
          B7-H5, B7H5, C10orf54, DD1alpha, Dies1, GI24, PD-1H, PP2135, SISP1, VISTA
          Chromosome Location:
          Chr.10: 71747556 - 71773520 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00735290_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: VSIR
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022153.2 NP_071436.1 5 - 6 786 95
          GenBank mRNA AF289594.1 - 1282 95
          AK024449.1 - 5 - 6 764 95
          AY265805.1 - 5 - 6 761 95
          AY358379.1 - 5 - 6 846 95
          BC020568.1 - 5 - 6 718 95
          BC111048.1 - 5 - 6 694 95
          BC127257.1 - 2 - 3 124 95
          EF506490.1 - 5 - 6 758 95

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1000010956.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC10001373.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene VSIR

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of gene expression
          positive regulation of endopeptidase activity
          positive regulation of cell migration
          zymogen activation
          negative regulation of interferon-gamma production
          negative regulation of interleukin-10 production
          negative regulation of interleukin-17 production
          negative regulation of tumor necrosis factor production
          positive regulation of regulatory T cell differentiation
          negative regulation of alpha-beta T cell activation
          regulation of immune response
          positive regulation of collagen catabolic process
          negative regulation of CD4-positive, alpha-beta T cell proliferation
          negative regulation of CD8-positive, alpha-beta T cell proliferation
          protein binding
          enzyme binding
          identical protein binding
          endopeptidase activator activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR10017364.hg.1 ENST00000263569.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10017364.hg.1 ENST00000394957.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10017364.hg.1 ENST00000470317.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10017364.hg.1 NM_022153.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10017364.hg.1 TC10001373.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1000010956.hg.1 ENST00000394957.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010956.hg.1 ENST00000394957.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010956.hg.1 ENST00000394957.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010956.hg.1 ENST00000394957.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010956.hg.1 ENST00000394957.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010956.hg.1 ENST00000394957.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010956.hg.1 ENST00000394957.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010956.hg.1 ENST00000394957.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010956.hg.1 ENST00000394957.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010956.hg.1 ENST00000394957.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010956.hg.1 ENST00000394957.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010956.hg.1 ENST00000394957.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010956.hg.1 ENST00000470317.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010956.hg.1 ENST00000470317.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010956.hg.1 ENST00000470317.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010956.hg.1 ENST00000470317.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010956.hg.1 ENST00000470317.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010956.hg.1 ENST00000470317.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010956.hg.1 ENST00000481568.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010956.hg.1 ENST00000481568.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010956.hg.1 ENST00000481568.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010956.hg.1 NM_022153.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010956.hg.1 NM_022153.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010956.hg.1 NM_022153.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010956.hg.1 NM_022153.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010956.hg.1 NM_022153.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010956.hg.1 NM_022153.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010956.hg.1 NM_022153.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010956.hg.1 NM_022153.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010956.hg.1 NM_022153.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010956.hg.1 NM_022153.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010956.hg.1 NM_022153.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010956.hg.1 NM_022153.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010956.hg.1 NM_022153.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000174519.hg.1 ENST00000394957.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1000174519.hg.1 ENST00000470317.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1000174519.hg.1 NM_022153.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000174519.hg.1 ENST00000394957.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1000174519.hg.1 ENST00000470317.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1000174519.hg.1 NM_022153.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000174519.hg.1 TC1000010956.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC1000010956.hg.1 ENST00000394957.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000010956.hg.1 ENST00000394957.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000010956.hg.1 ENST00000394957.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000010956.hg.1 ENST00000394957.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000010956.hg.1 ENST00000394957.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000010956.hg.1 ENST00000394957.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000010956.hg.1 ENST00000394957.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000010956.hg.1 ENST00000394957.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000010956.hg.1 ENST00000394957.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000010956.hg.1 ENST00000394957.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000010956.hg.1 ENST00000394957.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000010956.hg.1 ENST00000394957.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000010956.hg.1 ENST00000470317.2 Ensembl
          Affymetrix Human Clariom D Assay TC1000010956.hg.1 ENST00000470317.2 Ensembl
          Affymetrix Human Clariom D Assay TC1000010956.hg.1 ENST00000470317.2 Ensembl
          Affymetrix Human Clariom D Assay TC1000010956.hg.1 ENST00000470317.2 Ensembl
          Affymetrix Human Clariom D Assay TC1000010956.hg.1 ENST00000470317.2 Ensembl
          Affymetrix Human Clariom D Assay TC1000010956.hg.1 ENST00000470317.2 Ensembl
          Affymetrix Human Clariom D Assay TC1000010956.hg.1 ENST00000481568.2 Ensembl
          Affymetrix Human Clariom D Assay TC1000010956.hg.1 ENST00000481568.2 Ensembl
          Affymetrix Human Clariom D Assay TC1000010956.hg.1 ENST00000481568.2 Ensembl
          Affymetrix Human Clariom D Assay TC1000010956.hg.1 NM_022153.1 Refseq
          Affymetrix Human Clariom D Assay TC1000010956.hg.1 NM_022153.1 Refseq
          Affymetrix Human Clariom D Assay TC1000010956.hg.1 NM_022153.1 Refseq
          Affymetrix Human Clariom D Assay TC1000010956.hg.1 NM_022153.1 Refseq
          Affymetrix Human Clariom D Assay TC1000010956.hg.1 NM_022153.1 Refseq
          Affymetrix Human Clariom D Assay TC1000010956.hg.1 NM_022153.1 Refseq
          Affymetrix Human Clariom D Assay TC1000010956.hg.1 NM_022153.1 Refseq
          Affymetrix Human Clariom D Assay TC1000010956.hg.1 NM_022153.1 Refseq
          Affymetrix Human Clariom D Assay TC1000010956.hg.1 NM_022153.1 Refseq
          Affymetrix Human Clariom D Assay TC1000010956.hg.1 NM_022153.1 Refseq
          Affymetrix Human Clariom D Assay TC1000010956.hg.1 NM_022153.1 Refseq
          Affymetrix Human Clariom D Assay TC1000010956.hg.1 NM_022153.1 Refseq
          Affymetrix Human Clariom D Assay TC1000010956.hg.1 NM_022153.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline