| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010721.hg.1 |
ENST00000446432.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09017717.hg.1 |
ENST00000374865.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09017717.hg.1 |
NM_019051.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09017717.hg.1 |
TC09001420.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09017719.hg.1 |
ENST00000374865.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09017719.hg.1 |
ENST00000539624.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09017719.hg.1 |
NM_019051.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09017719.hg.1 |
TC09001420.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09017718.hg.1 |
ENST00000374865.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09017718.hg.1 |
ENST00000539624.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09017718.hg.1 |
NM_019051.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09017718.hg.1 |
TC09001420.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012243.hg.1 |
ENST00000431951.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015472.hg.1 |
ENST00000416821.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011031.hg.1 |
ENST00000374865.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011031.hg.1 |
ENST00000374865.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011031.hg.1 |
ENST00000374865.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011031.hg.1 |
ENST00000374865.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011031.hg.1 |
NM_019051.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011031.hg.1 |
NM_019051.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011031.hg.1 |
NM_019051.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009658.hg.1 |
ENST00000604356.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900175183.hg.1 |
ENST00000374865.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900175183.hg.1 |
NM_019051.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900175183.hg.1 |
ENST00000374865.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900175183.hg.1 |
NM_019051.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900175183.hg.1 |
TC0900011031.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0900175182.hg.1 |
ENST00000374865.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900175182.hg.1 |
NM_019051.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900175182.hg.1 |
ENST00000374865.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900175182.hg.1 |
NM_019051.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900175182.hg.1 |
TC0900011031.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0500009658.hg.1 |
ENST00000604356.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012243.hg.1 |
ENST00000431951.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015472.hg.1 |
ENST00000416821.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011031.hg.1 |
ENST00000374865.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011031.hg.1 |
ENST00000374865.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011031.hg.1 |
ENST00000374865.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011031.hg.1 |
ENST00000374865.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011031.hg.1 |
NM_019051.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011031.hg.1 |
NM_019051.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011031.hg.1 |
NM_019051.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000010721.hg.1 |
ENST00000446432.1 |
Ensembl |