Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00907826_m1
          See other TAF5 GE Assays ›
          Gene Symbol:
          TAF5
          Gene Name:
          TATA-box binding protein associated factor 5
          Gene Aliases:
          TAF(II)100, TAF2D, TAFII-100, TAFII100
          Chromosome Location:
          Chr.10: 103367953 - 103389065 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00907826_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: TAF5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_006951.4 NP_008882.2 8 - 9 1866 123
          GenBank mRNA AK223615.1 - 8 - 9 1838 123
          AK291229.1 - 8 - 9 1866 123
          BC052268.1 - 8 - 9 1844 6575596 123
          BC136348.1 - 8 - 9 1847 9020335 123
          U75309.1 - 8 - 9 1832 123
          U80191.1 - 8 - 9 1843 123
          X95525.1 - 8 - 9 1852 123

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1000008769.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC10000772.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene TAF5

          Gene Ontology Categories:

          Function(s) Process(es)

          DNA-templated transcription, initiation
          regulation of transcription, DNA-templated
          transcription from RNA polymerase II promoter
          transcription initiation from RNA polymerase II promoter
          transcription elongation from RNA polymerase II promoter
          viral process
          chromatin modification
          histone acetylation
          snRNA transcription from RNA polymerase II promoter
          regulation of signal transduction by p53 class mediator
          transcription factor activity, sequence-specific DNA binding
          histone acetyltransferase activity
          protein binding
          transcription regulatory region DNA binding
          protein dimerization activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1000008769.hg.1 ENST00000369839.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008769.hg.1 ENST00000369839.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008769.hg.1 ENST00000369839.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008769.hg.1 ENST00000369839.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008769.hg.1 ENST00000369839.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008769.hg.1 ENST00000369839.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008769.hg.1 ENST00000369839.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008769.hg.1 ENST00000369839.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008769.hg.1 ENST00000369839.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008769.hg.1 ENST00000369839.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008769.hg.1 ENST00000369839.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008769.hg.1 ENST00000369839.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008769.hg.1 ENST00000369839.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008769.hg.1 ENST00000369839.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008769.hg.1 NM_006951.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008769.hg.1 NM_006951.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008769.hg.1 NM_006951.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008769.hg.1 NM_006951.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008769.hg.1 NM_006951.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008769.hg.1 NM_006951.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008769.hg.1 NM_006951.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008769.hg.1 NM_006951.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008769.hg.1 NM_006951.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008769.hg.1 NM_006951.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008769.hg.1 NM_006951.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008769.hg.1 NM_006951.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008769.hg.1 NM_006951.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008769.hg.1 NM_006951.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10009188.hg.1 ENST00000351396.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10009188.hg.1 ENST00000369839.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10009188.hg.1 NM_006951.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10009188.hg.1 TC10000772.hg.1 HTA TC
          Affymetrix Human Clariom D Assay TC1000008769.hg.1 ENST00000369839.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000008769.hg.1 ENST00000369839.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000008769.hg.1 ENST00000369839.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000008769.hg.1 ENST00000369839.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000008769.hg.1 ENST00000369839.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000008769.hg.1 ENST00000369839.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000008769.hg.1 ENST00000369839.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000008769.hg.1 ENST00000369839.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000008769.hg.1 ENST00000369839.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000008769.hg.1 ENST00000369839.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000008769.hg.1 ENST00000369839.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000008769.hg.1 ENST00000369839.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000008769.hg.1 ENST00000369839.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000008769.hg.1 ENST00000369839.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000008769.hg.1 NM_006951.4 Refseq
          Affymetrix Human Clariom D Assay TC1000008769.hg.1 NM_006951.4 Refseq
          Affymetrix Human Clariom D Assay TC1000008769.hg.1 NM_006951.4 Refseq
          Affymetrix Human Clariom D Assay TC1000008769.hg.1 NM_006951.4 Refseq
          Affymetrix Human Clariom D Assay TC1000008769.hg.1 NM_006951.4 Refseq
          Affymetrix Human Clariom D Assay TC1000008769.hg.1 NM_006951.4 Refseq
          Affymetrix Human Clariom D Assay TC1000008769.hg.1 NM_006951.4 Refseq
          Affymetrix Human Clariom D Assay TC1000008769.hg.1 NM_006951.4 Refseq
          Affymetrix Human Clariom D Assay TC1000008769.hg.1 NM_006951.4 Refseq
          Affymetrix Human Clariom D Assay TC1000008769.hg.1 NM_006951.4 Refseq
          Affymetrix Human Clariom D Assay TC1000008769.hg.1 NM_006951.4 Refseq
          Affymetrix Human Clariom D Assay TC1000008769.hg.1 NM_006951.4 Refseq
          Affymetrix Human Clariom D Assay TC1000008769.hg.1 NM_006951.4 Refseq
          Affymetrix Human Clariom D Assay TC1000008769.hg.1 NM_006951.4 Refseq
          Affymetrix Human Clariom D Assay PSR1000160027.hg.1 ENST00000369839.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1000160027.hg.1 NM_006951.4 Refseq
          Affymetrix Human Clariom D Assay PSR1000160027.hg.1 ENST00000369839.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1000160027.hg.1 NM_006951.4 Refseq
          Affymetrix Human Clariom D Assay PSR1000160027.hg.1 TC1000008769.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline