Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00915026_m1
          See other PRPF6 GE Assays ›
          Gene Symbol:
          PRPF6
          Gene Name:
          pre-mRNA processing factor 6
          Gene Aliases:
          ANT-1, ANT1, C20orf14, Prp6, RP60, SNRNP102, TOM, U5-102K, hPrp6
          Chromosome Location:
          Chr.20: 63981069 - 64033100 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00915026_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PRPF6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012469.3 NP_036601.2 10 - 11 1473 117
          GenBank mRNA AB019219.1 - 10 - 11 1387 117
          AF026031.1 - 10 - 11 1404 117
          AF221842.1 - 10 - 11 1410 117
          AK001554.1 - 10 - 11 1416 117
          AK225993.1 - 10 - 11 1427 117
          AK314310.1 - 10 - 11 1397 117
          AL137320.1 - 1 - 2 104 117
          BC001666.2 - 10 - 11 1351 2988165 117

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC2000008104.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC20000531.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene PRPF6

          Panther Classification:

          Molecular Function -

          RNA splicing factor

          Gene Ontology Categories:

          Function(s) Process(es)

          spliceosomal tri-snRNP complex assembly
          spliceosomal complex assembly
          RNA splicing, via transesterification reactions
          mRNA splicing, via spliceosome
          RNA localization
          RNA splicing
          positive regulation of transcription from RNA polymerase II promoter
          transcription coactivator activity
          RNA binding
          protein binding
          ribonucleoprotein complex binding
          poly(A) RNA binding
          androgen receptor binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC2000008104.hg.1 ENST00000266079.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008104.hg.1 ENST00000266079.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008104.hg.1 ENST00000266079.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008104.hg.1 ENST00000266079.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008104.hg.1 ENST00000266079.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008104.hg.1 ENST00000266079.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008104.hg.1 ENST00000266079.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008104.hg.1 ENST00000266079.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008104.hg.1 ENST00000266079.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008104.hg.1 ENST00000266079.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008104.hg.1 ENST00000266079.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008104.hg.1 ENST00000266079.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008104.hg.1 ENST00000266079.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008104.hg.1 ENST00000266079.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008104.hg.1 ENST00000266079.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008104.hg.1 ENST00000266079.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008104.hg.1 ENST00000266079.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008104.hg.1 ENST00000266079.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008104.hg.1 ENST00000266079.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008104.hg.1 ENST00000266079.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008104.hg.1 ENST00000266079.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008104.hg.1 ENST00000266079.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008104.hg.1 ENST00000266079.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008104.hg.1 ENST00000266079.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008104.hg.1 NM_012469.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000008104.hg.1 NM_012469.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000008104.hg.1 NM_012469.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000008104.hg.1 NM_012469.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000008104.hg.1 NM_012469.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000008104.hg.1 NM_012469.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000008104.hg.1 NM_012469.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000008104.hg.1 NM_012469.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000008104.hg.1 NM_012469.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000008104.hg.1 NM_012469.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000008104.hg.1 NM_012469.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000008104.hg.1 NM_012469.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000008104.hg.1 NM_012469.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000008104.hg.1 NM_012469.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000008104.hg.1 NM_012469.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000008104.hg.1 NM_012469.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000008104.hg.1 NM_012469.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000008104.hg.1 NM_012469.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000008104.hg.1 NM_012469.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000008104.hg.1 NM_012469.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000008104.hg.1 NM_012469.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000008104.hg.1 NM_012469.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000008104.hg.1 NM_012469.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000008104.hg.1 NM_012469.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000008104.hg.1 NM_012469.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR20007594.hg.1 ENST00000266079.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR20007594.hg.1 ENST00000535781.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR20007594.hg.1 NM_012469.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR20007594.hg.1 TC20000531.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR2000156787.hg.1 ENST00000266079.4 Ensembl
          Affymetrix Human Clariom D Assay PSR2000156787.hg.1 NM_012469.3 Refseq
          Affymetrix Human Clariom D Assay PSR2000156787.hg.1 TC2000008104.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2000156787.hg.1 ENST00000266079.4 Ensembl
          Affymetrix Human Clariom D Assay PSR2000156787.hg.1 NM_012469.3 Refseq
          Affymetrix Human Clariom D Assay TC2000008104.hg.1 ENST00000266079.4 Ensembl
          Affymetrix Human Clariom D Assay TC2000008104.hg.1 ENST00000266079.4 Ensembl
          Affymetrix Human Clariom D Assay TC2000008104.hg.1 ENST00000266079.4 Ensembl
          Affymetrix Human Clariom D Assay TC2000008104.hg.1 ENST00000266079.4 Ensembl
          Affymetrix Human Clariom D Assay TC2000008104.hg.1 ENST00000266079.4 Ensembl
          Affymetrix Human Clariom D Assay TC2000008104.hg.1 ENST00000266079.4 Ensembl
          Affymetrix Human Clariom D Assay TC2000008104.hg.1 ENST00000266079.4 Ensembl
          Affymetrix Human Clariom D Assay TC2000008104.hg.1 ENST00000266079.4 Ensembl
          Affymetrix Human Clariom D Assay TC2000008104.hg.1 ENST00000266079.4 Ensembl
          Affymetrix Human Clariom D Assay TC2000008104.hg.1 ENST00000266079.4 Ensembl
          Affymetrix Human Clariom D Assay TC2000008104.hg.1 ENST00000266079.4 Ensembl
          Affymetrix Human Clariom D Assay TC2000008104.hg.1 ENST00000266079.4 Ensembl
          Affymetrix Human Clariom D Assay TC2000008104.hg.1 ENST00000266079.4 Ensembl
          Affymetrix Human Clariom D Assay TC2000008104.hg.1 ENST00000266079.4 Ensembl
          Affymetrix Human Clariom D Assay TC2000008104.hg.1 ENST00000266079.4 Ensembl
          Affymetrix Human Clariom D Assay TC2000008104.hg.1 ENST00000266079.4 Ensembl
          Affymetrix Human Clariom D Assay TC2000008104.hg.1 ENST00000266079.4 Ensembl
          Affymetrix Human Clariom D Assay TC2000008104.hg.1 ENST00000266079.4 Ensembl
          Affymetrix Human Clariom D Assay TC2000008104.hg.1 ENST00000266079.4 Ensembl
          Affymetrix Human Clariom D Assay TC2000008104.hg.1 ENST00000266079.4 Ensembl
          Affymetrix Human Clariom D Assay TC2000008104.hg.1 ENST00000266079.4 Ensembl
          Affymetrix Human Clariom D Assay TC2000008104.hg.1 ENST00000266079.4 Ensembl
          Affymetrix Human Clariom D Assay TC2000008104.hg.1 ENST00000266079.4 Ensembl
          Affymetrix Human Clariom D Assay TC2000008104.hg.1 ENST00000266079.4 Ensembl
          Affymetrix Human Clariom D Assay TC2000008104.hg.1 NM_012469.3 Refseq
          Affymetrix Human Clariom D Assay TC2000008104.hg.1 NM_012469.3 Refseq
          Affymetrix Human Clariom D Assay TC2000008104.hg.1 NM_012469.3 Refseq
          Affymetrix Human Clariom D Assay TC2000008104.hg.1 NM_012469.3 Refseq
          Affymetrix Human Clariom D Assay TC2000008104.hg.1 NM_012469.3 Refseq
          Affymetrix Human Clariom D Assay TC2000008104.hg.1 NM_012469.3 Refseq
          Affymetrix Human Clariom D Assay TC2000008104.hg.1 NM_012469.3 Refseq
          Affymetrix Human Clariom D Assay TC2000008104.hg.1 NM_012469.3 Refseq
          Affymetrix Human Clariom D Assay TC2000008104.hg.1 NM_012469.3 Refseq
          Affymetrix Human Clariom D Assay TC2000008104.hg.1 NM_012469.3 Refseq
          Affymetrix Human Clariom D Assay TC2000008104.hg.1 NM_012469.3 Refseq
          Affymetrix Human Clariom D Assay TC2000008104.hg.1 NM_012469.3 Refseq
          Affymetrix Human Clariom D Assay TC2000008104.hg.1 NM_012469.3 Refseq
          Affymetrix Human Clariom D Assay TC2000008104.hg.1 NM_012469.3 Refseq
          Affymetrix Human Clariom D Assay TC2000008104.hg.1 NM_012469.3 Refseq
          Affymetrix Human Clariom D Assay TC2000008104.hg.1 NM_012469.3 Refseq
          Affymetrix Human Clariom D Assay TC2000008104.hg.1 NM_012469.3 Refseq
          Affymetrix Human Clariom D Assay TC2000008104.hg.1 NM_012469.3 Refseq
          Affymetrix Human Clariom D Assay TC2000008104.hg.1 NM_012469.3 Refseq
          Affymetrix Human Clariom D Assay TC2000008104.hg.1 NM_012469.3 Refseq
          Affymetrix Human Clariom D Assay TC2000008104.hg.1 NM_012469.3 Refseq
          Affymetrix Human Clariom D Assay TC2000008104.hg.1 NM_012469.3 Refseq
          Affymetrix Human Clariom D Assay TC2000008104.hg.1 NM_012469.3 Refseq
          Affymetrix Human Clariom D Assay TC2000008104.hg.1 NM_012469.3 Refseq
          Affymetrix Human Clariom D Assay TC2000008104.hg.1 NM_012469.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline