Search
Search

GSDMA| Gene Symbol: GSDMA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178171.4 | NP_835465.2 | 8 - 9 | 869 | 78 | |
| XM_006721832.2 | XP_006721895.1 | 8 - 9 | 822 | 78 | ||
| XM_011524651.2 | XP_011522953.1 | 6 - 7 | 585 | 78 | ||
| GenBank mRNA | AB093591.1 | - | 7 - 8 | 762 | 78 | |
| AK096439.1 | - | 9 - 10 | 871 | 78 | ||
| BC109197.2 | - | 9 - 10 | 831 | 40005562 | 78 | |
Set Membership: |
|