Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00939741_g1
          See other TRAF6 GE Assays ›
          Gene Symbol:
          TRAF6
          Gene Name:
          TNF receptor associated factor 6
          Gene Aliases:
          MGC:3310, RNF85
          Chromosome Location:
          Chr.11: 36483767 - 36510313 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00939741_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: TRAF6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_004620.3 NP_004611.1 2 - 3 581 122
          NM_145803.2 NP_665802.1 3 - 4 674 122
          XM_017018220.1 XP_016873709.1 2 - 3 581 122
          GenBank mRNA AK292978.1 - 2 - 3 540 122
          BC031052.1 - 3 - 4 619 5272008 122
          U78798.1 - 2 - 3 514 122

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1100010551.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC11001560.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene TRAF6

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          activation of MAPK activity
          protein polyubiquitination
          ossification
          neural tube closure
          stimulatory C-type lectin receptor signaling pathway
          toll-like receptor signaling pathway
          positive regulation of T cell cytokine production
          MyD88-dependent toll-like receptor signaling pathway
          MyD88-independent toll-like receptor signaling pathway
          protein complex assembly
          I-kappaB kinase/NF-kappaB signaling
          activation of NF-kappaB-inducing kinase activity
          JNK cascade
          antigen processing and presentation of exogenous peptide antigen via MHC class II
          osteoclast differentiation
          membrane protein intracellular domain proteolysis
          positive regulation of protein ubiquitination
          positive regulation of lipopolysaccharide-mediated signaling pathway
          activation of protein kinase activity
          positive regulation of interleukin-2 production
          toll-like receptor 9 signaling pathway
          Fc-epsilon receptor signaling pathway
          T-helper 1 type immune response
          positive regulation of T cell proliferation
          odontogenesis of dentin-containing tooth
          myeloid dendritic cell differentiation
          positive regulation of apoptotic process
          negative regulation of apoptotic process
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          positive regulation of JUN kinase activity
          positive regulation of interleukin-12 biosynthetic process
          positive regulation of interleukin-6 biosynthetic process
          bone resorption
          positive regulation of osteoclast differentiation
          negative regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          cell development
          positive regulation of smooth muscle cell proliferation
          T cell receptor signaling pathway
          positive regulation of T cell activation
          regulation of immunoglobulin secretion
          positive regulation of sequence-specific DNA binding transcription factor activity
          positive regulation of NF-kappaB transcription factor activity
          protein autoubiquitination
          nucleotide-binding oligomerization domain containing signaling pathway
          interleukin-1-mediated signaling pathway
          protein K63-linked ubiquitination
          response to interleukin-1
          cellular response to lipopolysaccharide
          positive regulation of transcription regulatory region DNA binding
          ubiquitin-protein transferase activity
          tumor necrosis factor receptor binding
          protein binding
          zinc ion binding
          ligase activity
          protein kinase binding
          mitogen-activated protein kinase kinase kinase binding
          ubiquitin conjugating enzyme binding
          ubiquitin protein ligase binding
          thioesterase binding
          identical protein binding
          histone deacetylase binding
          protein kinase B binding
          protein N-terminus binding
          ubiquitin protein ligase activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR11019581.hg.1 ENST00000348124.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11019581.hg.1 ENST00000526995.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11019581.hg.1 NM_004620.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11019581.hg.1 NM_145803.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11019581.hg.1 TC11001560.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 ENST00000348124.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 ENST00000348124.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 ENST00000348124.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 ENST00000348124.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 ENST00000348124.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 ENST00000348124.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 ENST00000348124.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 ENST00000348124.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 ENST00000348124.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 ENST00000348124.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 ENST00000526995.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 ENST00000526995.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 ENST00000526995.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 ENST00000526995.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 ENST00000526995.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 ENST00000526995.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 ENST00000526995.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 ENST00000526995.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 ENST00000526995.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 ENST00000526995.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 ENST00000526995.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 ENST00000529150.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 ENST00000529150.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 ENST00000529150.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 ENST00000529150.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 NM_004620.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 NM_004620.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 NM_004620.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 NM_004620.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 NM_004620.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 NM_004620.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 NM_004620.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 NM_004620.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 NM_004620.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 NM_004620.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 NM_004620.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 NM_004620.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 NM_004620.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 NM_145803.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 NM_145803.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 NM_145803.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 NM_145803.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 NM_145803.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 NM_145803.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 NM_145803.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 NM_145803.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 NM_145803.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 NM_145803.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 NM_145803.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 NM_145803.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 NM_145803.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100010551.hg.1 NM_145803.2 Refseq
          Affymetrix Human Clariom D Assay PSR1100180087.hg.1 ENST00000348124.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1100180087.hg.1 ENST00000526995.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1100180087.hg.1 NM_004620.3 Refseq
          Affymetrix Human Clariom D Assay PSR1100180087.hg.1 NM_145803.2 Refseq
          Affymetrix Human Clariom D Assay PSR1100180087.hg.1 ENST00000348124.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1100180087.hg.1 ENST00000526995.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1100180087.hg.1 NM_004620.3 Refseq
          Affymetrix Human Clariom D Assay PSR1100180087.hg.1 NM_145803.2 Refseq
          Affymetrix Human Clariom D Assay PSR1100180087.hg.1 TC1100010551.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 ENST00000348124.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 ENST00000348124.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 ENST00000348124.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 ENST00000348124.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 ENST00000348124.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 ENST00000348124.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 ENST00000348124.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 ENST00000348124.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 ENST00000348124.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 ENST00000348124.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 ENST00000526995.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 ENST00000526995.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 ENST00000526995.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 ENST00000526995.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 ENST00000526995.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 ENST00000526995.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 ENST00000526995.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 ENST00000526995.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 ENST00000526995.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 ENST00000526995.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 ENST00000526995.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 ENST00000529150.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 ENST00000529150.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 ENST00000529150.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 ENST00000529150.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 NM_004620.3 Refseq
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 NM_004620.3 Refseq
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 NM_004620.3 Refseq
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 NM_004620.3 Refseq
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 NM_004620.3 Refseq
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 NM_004620.3 Refseq
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 NM_004620.3 Refseq
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 NM_004620.3 Refseq
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 NM_004620.3 Refseq
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 NM_004620.3 Refseq
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 NM_004620.3 Refseq
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 NM_004620.3 Refseq
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 NM_004620.3 Refseq
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 NM_145803.2 Refseq
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 NM_145803.2 Refseq
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 NM_145803.2 Refseq
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 NM_145803.2 Refseq
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 NM_145803.2 Refseq
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 NM_145803.2 Refseq
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 NM_145803.2 Refseq
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 NM_145803.2 Refseq
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 NM_145803.2 Refseq
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 NM_145803.2 Refseq
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 NM_145803.2 Refseq
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 NM_145803.2 Refseq
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 NM_145803.2 Refseq
          Affymetrix Human Clariom D Assay TC1100010551.hg.1 NM_145803.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline