Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01001478_m1
          See other ITGB3 GE Assays ›
          Gene Symbol:
          ITGB3
          Gene Name:
          integrin subunit beta 3
          Gene Aliases:
          BDPLT16, BDPLT2, CD61, GP3A, GPIIIa, GT
          Chromosome Location:
          Chr.17: 47253842 - 47312711 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01001478_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ITGB3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_000212.2 NP_000203.2 9 - 10 1277 70
          GenBank mRNA BC127666.1 - 9 - 10 1277 40128462 70
          BC127667.1 - 9 - 10 1281 40128464 70
          J02703.1 - 9 - 10 1277 70
          KC120777.1 - 9 - 10 1277 70
          KC120778.1 - 9 - 10 1277 70
          M20311.1 - 9 - 10 1273 70
          M35999.1 - 9 - 10 1275 70
          U95204.1 - 9 - 10 1257 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1700012274.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC17002878.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Human Extracellular Matrix and Adhesion Molecules Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene ITGB3

          Panther Classification:

          Molecular Function -

          integrin

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of protein phosphorylation
          positive regulation of endothelial cell proliferation
          platelet degranulation
          cell-substrate junction assembly
          cell adhesion
          cell-matrix adhesion
          integrin-mediated signaling pathway
          blood coagulation
          positive regulation of endothelial cell migration
          negative regulation of macrophage derived foam cell differentiation
          negative regulation of lipid storage
          smooth muscle cell migration
          cell growth
          cell migration
          platelet activation
          extracellular matrix organization
          positive regulation of vascular endothelial growth factor receptor signaling pathway
          cell-substrate adhesion
          activation of protein kinase activity
          negative regulation of lipid transport
          heterotypic cell-cell adhesion
          substrate adhesion-dependent cell spreading
          tube development
          apolipoprotein A-I-mediated signaling pathway
          wound healing
          regulation of bone resorption
          negative regulation of low-density lipoprotein particle receptor biosynthetic process
          viral entry into host cell
          vascular endothelial growth factor receptor signaling pathway
          mesodermal cell differentiation
          positive regulation of peptidyl-tyrosine phosphorylation
          negative regulation of lipoprotein metabolic process
          leukocyte migration
          negative chemotaxis
          angiogenesis involved in wound healing
          platelet aggregation
          virus receptor activity
          fibronectin binding
          protease binding
          protein disulfide isomerase activity
          platelet-derived growth factor receptor binding
          protein binding
          coreceptor activity
          fibroblast growth factor binding
          enzyme binding
          C-X3-C chemokine binding
          insulin-like growth factor I binding
          neuregulin binding
          identical protein binding
          vascular endothelial growth factor receptor 2 binding
          cell adhesion molecule binding
          extracellular matrix binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 ENST00000559488.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 ENST00000559488.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 ENST00000559488.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 ENST00000559488.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 ENST00000559488.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 ENST00000559488.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 ENST00000559488.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 ENST00000559488.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 ENST00000559488.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 ENST00000559488.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 ENST00000559488.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 ENST00000559488.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 ENST00000559488.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 ENST00000559488.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 ENST00000559488.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 ENST00000559488.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 ENST00000559488.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 ENST00000559488.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 ENST00000559488.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 ENST00000571680.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 ENST00000571680.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 ENST00000571680.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 ENST00000571680.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 ENST00000571680.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 ENST00000571680.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 ENST00000571680.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 ENST00000571680.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 ENST00000571680.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 ENST00000571680.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 ENST00000571680.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 ENST00000571680.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 ENST00000571680.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 ENST00000573377.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 ENST00000573377.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 ENST00000573377.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 ENST00000573377.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 NM_000212.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 NM_000212.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 NM_000212.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 NM_000212.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 NM_000212.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 NM_000212.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 NM_000212.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 NM_000212.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 NM_000212.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 NM_000212.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 NM_000212.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 NM_000212.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 NM_000212.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 NM_000212.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 NM_000212.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 NM_000212.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 NM_000212.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 NM_000212.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 NM_000212.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 NM_000212.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17007426.hg.1 ENST00000262017.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17007426.hg.1 ENST00000559488.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17007426.hg.1 NM_000212.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17007426.hg.1 TC17002878.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1700202756.hg.1 ENST00000559488.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1700202756.hg.1 ENST00000571680.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700202756.hg.1 ENST00000573377.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700202756.hg.1 NM_000212.2 Refseq
          Affymetrix Human Clariom D Assay PSR1700202756.hg.1 TC1700012274.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1700202756.hg.1 ENST00000559488.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1700202756.hg.1 ENST00000571680.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700202756.hg.1 ENST00000573377.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700202756.hg.1 NM_000212.2 Refseq
          Affymetrix Human Clariom D Assay PSR1700202760.hg.1 ENST00000559488.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1700202760.hg.1 ENST00000573377.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700202760.hg.1 NM_000212.2 Refseq
          Affymetrix Human Clariom D Assay PSR1700202760.hg.1 TC1700012274.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1700202760.hg.1 ENST00000559488.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1700202760.hg.1 ENST00000573377.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700202760.hg.1 NM_000212.2 Refseq
          Affymetrix Human Clariom D Assay PSR1700202761.hg.1 ENST00000559488.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1700202761.hg.1 NM_000212.2 Refseq
          Affymetrix Human Clariom D Assay PSR1700202761.hg.1 TC1700012274.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1700202761.hg.1 ENST00000559488.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1700202761.hg.1 NM_000212.2 Refseq
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 ENST00000559488.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 ENST00000559488.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 ENST00000559488.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 ENST00000559488.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 ENST00000559488.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 ENST00000559488.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 ENST00000559488.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 ENST00000559488.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 ENST00000559488.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 ENST00000559488.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 ENST00000559488.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 ENST00000559488.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 ENST00000559488.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 ENST00000559488.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 ENST00000559488.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 ENST00000559488.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 ENST00000559488.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 ENST00000559488.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 ENST00000559488.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 ENST00000571680.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 ENST00000571680.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 ENST00000571680.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 ENST00000571680.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 ENST00000571680.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 ENST00000571680.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 ENST00000571680.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 ENST00000571680.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 ENST00000571680.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 ENST00000571680.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 ENST00000571680.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 ENST00000571680.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 ENST00000571680.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 ENST00000573377.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 ENST00000573377.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 ENST00000573377.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 ENST00000573377.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 NM_000212.2 Refseq
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 NM_000212.2 Refseq
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 NM_000212.2 Refseq
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 NM_000212.2 Refseq
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 NM_000212.2 Refseq
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 NM_000212.2 Refseq
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 NM_000212.2 Refseq
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 NM_000212.2 Refseq
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 NM_000212.2 Refseq
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 NM_000212.2 Refseq
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 NM_000212.2 Refseq
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 NM_000212.2 Refseq
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 NM_000212.2 Refseq
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 NM_000212.2 Refseq
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 NM_000212.2 Refseq
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 NM_000212.2 Refseq
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 NM_000212.2 Refseq
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 NM_000212.2 Refseq
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 NM_000212.2 Refseq
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 NM_000212.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline