| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010209.hg.1 |
ENST00000292090.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010209.hg.1 |
ENST00000292090.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010209.hg.1 |
ENST00000292090.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010209.hg.1 |
ENST00000292090.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010209.hg.1 |
ENST00000292090.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010209.hg.1 |
ENST00000292090.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010209.hg.1 |
ENST00000292090.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010209.hg.1 |
ENST00000292090.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010209.hg.1 |
ENST00000292090.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010209.hg.1 |
ENST00000292090.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010209.hg.1 |
ENST00000292090.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010209.hg.1 |
ENST00000394933.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010209.hg.1 |
ENST00000394933.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010209.hg.1 |
ENST00000394933.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010209.hg.1 |
ENST00000394933.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010209.hg.1 |
ENST00000394933.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010209.hg.1 |
ENST00000394933.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010209.hg.1 |
ENST00000394933.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010209.hg.1 |
ENST00000394933.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010209.hg.1 |
ENST00000394933.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010209.hg.1 |
ENST00000394933.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010209.hg.1 |
ENST00000580518.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010209.hg.1 |
ENST00000580518.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010209.hg.1 |
ENST00000580518.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010209.hg.1 |
ENST00000580518.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010209.hg.1 |
ENST00000580518.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010209.hg.1 |
ENST00000580518.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010209.hg.1 |
ENST00000580518.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010209.hg.1 |
ENST00000581236.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010209.hg.1 |
ENST00000581236.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010209.hg.1 |
ENST00000581236.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010209.hg.1 |
ENST00000581236.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010209.hg.1 |
ENST00000581236.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010209.hg.1 |
NM_001160407.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010209.hg.1 |
NM_001160407.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010209.hg.1 |
NM_001160407.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010209.hg.1 |
NM_001160407.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010209.hg.1 |
NM_001160407.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010209.hg.1 |
NM_001160407.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010209.hg.1 |
NM_001160407.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010209.hg.1 |
NM_001160407.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010209.hg.1 |
NM_001160407.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010209.hg.1 |
NM_001160407.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010209.hg.1 |
NM_001160407.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010209.hg.1 |
NM_138463.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010209.hg.1 |
NM_138463.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010209.hg.1 |
NM_138463.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010209.hg.1 |
NM_138463.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010209.hg.1 |
NM_138463.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010209.hg.1 |
NM_138463.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010209.hg.1 |
NM_138463.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010209.hg.1 |
NM_138463.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010209.hg.1 |
NM_138463.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010209.hg.1 |
NM_138463.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010209.hg.1 |
NM_138463.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17017448.hg.1 |
ENST00000292090.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17017448.hg.1 |
ENST00000394933.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17017448.hg.1 |
NM_001160407.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17017448.hg.1 |
NM_138463.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17017448.hg.1 |
TC17001306.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC1700010209.hg.1 |
ENST00000292090.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010209.hg.1 |
ENST00000292090.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010209.hg.1 |
ENST00000292090.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010209.hg.1 |
ENST00000292090.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010209.hg.1 |
ENST00000292090.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010209.hg.1 |
ENST00000292090.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010209.hg.1 |
ENST00000292090.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010209.hg.1 |
ENST00000292090.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010209.hg.1 |
ENST00000292090.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010209.hg.1 |
ENST00000292090.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010209.hg.1 |
ENST00000292090.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010209.hg.1 |
ENST00000394933.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010209.hg.1 |
ENST00000394933.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010209.hg.1 |
ENST00000394933.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010209.hg.1 |
ENST00000394933.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010209.hg.1 |
ENST00000394933.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010209.hg.1 |
ENST00000394933.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010209.hg.1 |
ENST00000394933.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010209.hg.1 |
ENST00000394933.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010209.hg.1 |
ENST00000394933.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010209.hg.1 |
ENST00000394933.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010209.hg.1 |
ENST00000580518.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010209.hg.1 |
ENST00000580518.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010209.hg.1 |
ENST00000580518.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010209.hg.1 |
ENST00000580518.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010209.hg.1 |
ENST00000580518.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010209.hg.1 |
ENST00000580518.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010209.hg.1 |
ENST00000580518.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010209.hg.1 |
ENST00000581236.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010209.hg.1 |
ENST00000581236.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010209.hg.1 |
ENST00000581236.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010209.hg.1 |
ENST00000581236.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010209.hg.1 |
ENST00000581236.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010209.hg.1 |
NM_001160407.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010209.hg.1 |
NM_001160407.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010209.hg.1 |
NM_001160407.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010209.hg.1 |
NM_001160407.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010209.hg.1 |
NM_001160407.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010209.hg.1 |
NM_001160407.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010209.hg.1 |
NM_001160407.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010209.hg.1 |
NM_001160407.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010209.hg.1 |
NM_001160407.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010209.hg.1 |
NM_001160407.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010209.hg.1 |
NM_001160407.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010209.hg.1 |
NM_138463.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010209.hg.1 |
NM_138463.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010209.hg.1 |
NM_138463.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010209.hg.1 |
NM_138463.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010209.hg.1 |
NM_138463.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010209.hg.1 |
NM_138463.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010209.hg.1 |
NM_138463.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010209.hg.1 |
NM_138463.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010209.hg.1 |
NM_138463.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010209.hg.1 |
NM_138463.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010209.hg.1 |
NM_138463.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700180199.hg.1 |
ENST00000292090.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700180199.hg.1 |
ENST00000394933.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700180199.hg.1 |
ENST00000580518.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700180199.hg.1 |
NM_001160407.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700180199.hg.1 |
NM_138463.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700180199.hg.1 |
TC1700010209.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1700180199.hg.1 |
ENST00000292090.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700180199.hg.1 |
ENST00000394933.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700180199.hg.1 |
ENST00000580518.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700180199.hg.1 |
NM_001160407.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700180199.hg.1 |
NM_138463.3 |
Refseq |