| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01001610.hg.1 |
ENST00000361923.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01001610.hg.1 |
ENST00000377532.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01001610.hg.1 |
ENST00000377541.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01001610.hg.1 |
ENST00000539773.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01001610.hg.1 |
NM_016831.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01001610.hg.1 |
TC01000100.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01001612.hg.1 |
ENST00000361923.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01001612.hg.1 |
ENST00000377532.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01001612.hg.1 |
ENST00000377541.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01001612.hg.1 |
ENST00000539773.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01001612.hg.1 |
NM_016831.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01001612.hg.1 |
TC01000100.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0100146579.hg.1 |
NM_001289864.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100146579.hg.1 |
NM_001289864.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100146579.hg.1 |
TC0100006725.hg.1 |
Clariom D TC |