Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01007892_g1
          See other KIF5A GE Assays ›
          Gene Symbol:
          KIF5A
          Gene Name:
          kinesin family member 5A
          Gene Aliases:
          D12S1889, MY050, NKHC, SPG10
          Chromosome Location:
          Chr.12: 57550064 - 57584771 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01007892_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: KIF5A
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_004984.2 NP_004975.2 25 - 26 3123 55
          GenBank mRNA AB210045.1 - 25 - 26 2948 55
          AB290174.1 - 25 - 26 3189 55
          AK294881.1 - 22 - 23 2876 55
          AK315202.1 - 25 - 26 3143 55
          BC146670.1 - 25 - 26 3126 40148192 55
          BC150208.1 - 25 - 26 3126 40148890 55
          BM930000.1 - 2 - 3 319 55
          U06698.1 - 25 - 26 3063 55

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1200012651.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC12000534.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene KIF5A

          Panther Classification:

          Molecular Function -

          microtubule binding motor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          retrograde vesicle-mediated transport, Golgi to ER
          microtubule-based movement
          chemical synaptic transmission
          axon guidance
          protein localization
          antigen processing and presentation of exogenous peptide antigen via MHC class II
          cytoskeleton-dependent intracellular transport
          motor activity
          protein binding
          ATP binding
          microtubule binding
          ATP-dependent microtubule motor activity, plus-end-directed

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR12007098.hg.1 ENST00000286452.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12007098.hg.1 ENST00000455537.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12007098.hg.1 NM_004984.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12007098.hg.1 TC12000534.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1200012651.hg.1 ENST00000286452.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012651.hg.1 ENST00000286452.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012651.hg.1 ENST00000286452.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012651.hg.1 ENST00000286452.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012651.hg.1 ENST00000286452.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012651.hg.1 ENST00000286452.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012651.hg.1 ENST00000286452.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012651.hg.1 ENST00000286452.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012651.hg.1 ENST00000286452.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012651.hg.1 ENST00000286452.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012651.hg.1 ENST00000286452.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012651.hg.1 ENST00000286452.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012651.hg.1 ENST00000286452.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012651.hg.1 ENST00000286452.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012651.hg.1 ENST00000286452.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012651.hg.1 ENST00000286452.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012651.hg.1 ENST00000286452.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012651.hg.1 ENST00000286452.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012651.hg.1 ENST00000286452.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012651.hg.1 ENST00000286452.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012651.hg.1 ENST00000286452.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012651.hg.1 ENST00000286452.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012651.hg.1 ENST00000286452.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012651.hg.1 ENST00000286452.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012651.hg.1 ENST00000286452.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012651.hg.1 ENST00000286452.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012651.hg.1 ENST00000286452.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012651.hg.1 ENST00000286452.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012651.hg.1 ENST00000286452.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012651.hg.1 ENST00000552227.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012651.hg.1 ENST00000552227.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012651.hg.1 ENST00000552227.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012651.hg.1 ENST00000552227.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012651.hg.1 NM_004984.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200012651.hg.1 NM_004984.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200012651.hg.1 NM_004984.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200012651.hg.1 NM_004984.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200012651.hg.1 NM_004984.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200012651.hg.1 NM_004984.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200012651.hg.1 NM_004984.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200012651.hg.1 NM_004984.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200012651.hg.1 NM_004984.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200012651.hg.1 NM_004984.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200012651.hg.1 NM_004984.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200012651.hg.1 NM_004984.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200012651.hg.1 NM_004984.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200012651.hg.1 NM_004984.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200012651.hg.1 NM_004984.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200012651.hg.1 NM_004984.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200012651.hg.1 NM_004984.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200012651.hg.1 NM_004984.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200012651.hg.1 NM_004984.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200012651.hg.1 NM_004984.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200012651.hg.1 NM_004984.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200012651.hg.1 NM_004984.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200012651.hg.1 NM_004984.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200012651.hg.1 NM_004984.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200012651.hg.1 NM_004984.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200012651.hg.1 NM_004984.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200012651.hg.1 NM_004984.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200012651.hg.1 NM_004984.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200012651.hg.1 NM_004984.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200012651.hg.1 NM_004984.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200012651.hg.1 NM_004984.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200012651.hg.1 NM_004984.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200012651.hg.1 NM_004984.3 Refseq
          Affymetrix Human Clariom D Assay TC1200012651.hg.1 ENST00000286452.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200012651.hg.1 ENST00000286452.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200012651.hg.1 ENST00000286452.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200012651.hg.1 ENST00000286452.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200012651.hg.1 ENST00000286452.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200012651.hg.1 ENST00000286452.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200012651.hg.1 ENST00000286452.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200012651.hg.1 ENST00000286452.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200012651.hg.1 ENST00000286452.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200012651.hg.1 ENST00000286452.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200012651.hg.1 ENST00000286452.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200012651.hg.1 ENST00000286452.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200012651.hg.1 ENST00000286452.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200012651.hg.1 ENST00000286452.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200012651.hg.1 ENST00000286452.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200012651.hg.1 ENST00000286452.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200012651.hg.1 ENST00000286452.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200012651.hg.1 ENST00000286452.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200012651.hg.1 ENST00000286452.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200012651.hg.1 ENST00000286452.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200012651.hg.1 ENST00000286452.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200012651.hg.1 ENST00000286452.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200012651.hg.1 ENST00000286452.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200012651.hg.1 ENST00000286452.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200012651.hg.1 ENST00000286452.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200012651.hg.1 ENST00000286452.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200012651.hg.1 ENST00000286452.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200012651.hg.1 ENST00000286452.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200012651.hg.1 ENST00000286452.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200012651.hg.1 ENST00000552227.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200012651.hg.1 ENST00000552227.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200012651.hg.1 ENST00000552227.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200012651.hg.1 ENST00000552227.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200012651.hg.1 NM_004984.3 Refseq
          Affymetrix Human Clariom D Assay TC1200012651.hg.1 NM_004984.3 Refseq
          Affymetrix Human Clariom D Assay TC1200012651.hg.1 NM_004984.3 Refseq
          Affymetrix Human Clariom D Assay TC1200012651.hg.1 NM_004984.3 Refseq
          Affymetrix Human Clariom D Assay TC1200012651.hg.1 NM_004984.3 Refseq
          Affymetrix Human Clariom D Assay TC1200012651.hg.1 NM_004984.3 Refseq
          Affymetrix Human Clariom D Assay TC1200012651.hg.1 NM_004984.3 Refseq
          Affymetrix Human Clariom D Assay TC1200012651.hg.1 NM_004984.3 Refseq
          Affymetrix Human Clariom D Assay TC1200012651.hg.1 NM_004984.3 Refseq
          Affymetrix Human Clariom D Assay TC1200012651.hg.1 NM_004984.3 Refseq
          Affymetrix Human Clariom D Assay TC1200012651.hg.1 NM_004984.3 Refseq
          Affymetrix Human Clariom D Assay TC1200012651.hg.1 NM_004984.3 Refseq
          Affymetrix Human Clariom D Assay TC1200012651.hg.1 NM_004984.3 Refseq
          Affymetrix Human Clariom D Assay TC1200012651.hg.1 NM_004984.3 Refseq
          Affymetrix Human Clariom D Assay TC1200012651.hg.1 NM_004984.3 Refseq
          Affymetrix Human Clariom D Assay TC1200012651.hg.1 NM_004984.3 Refseq
          Affymetrix Human Clariom D Assay TC1200012651.hg.1 NM_004984.3 Refseq
          Affymetrix Human Clariom D Assay TC1200012651.hg.1 NM_004984.3 Refseq
          Affymetrix Human Clariom D Assay TC1200012651.hg.1 NM_004984.3 Refseq
          Affymetrix Human Clariom D Assay TC1200012651.hg.1 NM_004984.3 Refseq
          Affymetrix Human Clariom D Assay TC1200012651.hg.1 NM_004984.3 Refseq
          Affymetrix Human Clariom D Assay TC1200012651.hg.1 NM_004984.3 Refseq
          Affymetrix Human Clariom D Assay TC1200012651.hg.1 NM_004984.3 Refseq
          Affymetrix Human Clariom D Assay TC1200012651.hg.1 NM_004984.3 Refseq
          Affymetrix Human Clariom D Assay TC1200012651.hg.1 NM_004984.3 Refseq
          Affymetrix Human Clariom D Assay TC1200012651.hg.1 NM_004984.3 Refseq
          Affymetrix Human Clariom D Assay TC1200012651.hg.1 NM_004984.3 Refseq
          Affymetrix Human Clariom D Assay TC1200012651.hg.1 NM_004984.3 Refseq
          Affymetrix Human Clariom D Assay TC1200012651.hg.1 NM_004984.3 Refseq
          Affymetrix Human Clariom D Assay TC1200012651.hg.1 NM_004984.3 Refseq
          Affymetrix Human Clariom D Assay TC1200012651.hg.1 NM_004984.3 Refseq
          Affymetrix Human Clariom D Assay TC1200012651.hg.1 NM_004984.3 Refseq
          Affymetrix Human Clariom D Assay TC1200012651.hg.1 NM_004984.3 Refseq
          Affymetrix Human Clariom D Assay PSR1200200747.hg.1 ENST00000286452.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200200747.hg.1 ENST00000552227.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200200747.hg.1 NM_004984.3 Refseq
          Affymetrix Human Clariom D Assay PSR1200200747.hg.1 ENST00000286452.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200200747.hg.1 ENST00000552227.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200200747.hg.1 NM_004984.3 Refseq
          Affymetrix Human Clariom D Assay PSR1200200747.hg.1 TC1200012651.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline