Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01018738_m1
          See other NR5A1 GE Assays ›
          Gene Symbol:
          NR5A1
          Gene Name:
          nuclear receptor subfamily 5 group A member 1
          Gene Aliases:
          AD4BP, ELP, FTZ1, FTZF1, POF7, SF-1, SF1, SPGF8, SRXX4, SRXY3, hSF-1
          Chromosome Location:
          Chr.9: 124481236 - 124507399 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01018738_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: NR5A1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_004959.5 NP_004950.2 4 - 5 1035 56
          GenBank mRNA AB307718.1 - 3 - 4 869 56
          BC032501.1 - 4 - 5 1056 56
          HQ692856.1 - 3 - 4 869 56
          U76388.1 - 3 - 4 884 56

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0900011499.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC09001579.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene NR5A1

          Gene Ontology Categories:

          Function(s) Process(es)

          luteinization
          regulation of transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          transcription from RNA polymerase II promoter
          sex determination
          primary sex determination
          male gonad development
          female gonad development
          hormone-mediated signaling pathway
          tissue development
          positive regulation of gene expression
          male sex determination
          adrenal gland development
          intracellular receptor signaling pathway
          Leydig cell differentiation
          hormone metabolic process
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          regulation of steroid biosynthetic process
          maintenance of protein location in nucleus
          Sertoli cell differentiation
          response to gonadotropin-releasing hormone
          calcineurin-mediated signaling
          positive regulation of male gonad development
          negative regulation of female gonad development
          RNA polymerase II regulatory region sequence-specific DNA binding
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          RNA polymerase II transcription factor activity, sequence-specific DNA binding
          transcription cofactor binding
          DNA binding
          chromatin binding
          double-stranded DNA binding
          transcription factor activity, sequence-specific DNA binding
          RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding
          protein binding
          phospholipid binding
          zinc ion binding
          lipid binding
          enzyme binding
          sequence-specific DNA binding
          metal ion binding
          sequence-specific double-stranded DNA binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0900011499.hg.1 ENST00000373587.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011499.hg.1 ENST00000373587.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011499.hg.1 ENST00000373587.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011499.hg.1 ENST00000373587.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011499.hg.1 ENST00000373587.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011499.hg.1 ENST00000373587.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011499.hg.1 ENST00000373588.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011499.hg.1 ENST00000373588.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011499.hg.1 ENST00000373588.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011499.hg.1 ENST00000373588.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011499.hg.1 ENST00000373588.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011499.hg.1 ENST00000373588.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011499.hg.1 ENST00000373588.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011499.hg.1 ENST00000373588.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011499.hg.1 ENST00000373588.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011499.hg.1 ENST00000373588.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011499.hg.1 ENST00000373588.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011499.hg.1 ENST00000455734.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011499.hg.1 ENST00000455734.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011499.hg.1 ENST00000455734.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011499.hg.1 ENST00000455734.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011499.hg.1 ENST00000620110.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011499.hg.1 ENST00000620110.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011499.hg.1 ENST00000620110.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011499.hg.1 ENST00000620110.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011499.hg.1 ENST00000620110.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011499.hg.1 ENST00000620110.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011499.hg.1 ENST00000620110.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011499.hg.1 ENST00000620110.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011499.hg.1 ENST00000620110.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011499.hg.1 ENST00000620110.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011499.hg.1 NM_004959.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900011499.hg.1 NM_004959.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900011499.hg.1 NM_004959.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900011499.hg.1 NM_004959.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900011499.hg.1 NM_004959.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900011499.hg.1 NM_004959.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900011499.hg.1 NM_004959.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900011499.hg.1 NM_004959.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900011499.hg.1 NM_004959.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900011499.hg.1 NM_004959.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900011499.hg.1 NM_004959.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR09019808.hg.1 ENST00000373588.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09019808.hg.1 NM_004959.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR09019808.hg.1 TC09001579.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR09019809.hg.1 ENST00000373588.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09019809.hg.1 NM_004959.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR09019809.hg.1 TC09001579.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0900178410.hg.1 ENST00000373587.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0900178410.hg.1 ENST00000373588.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0900178410.hg.1 NM_004959.4 Refseq
          Affymetrix Human Clariom D Assay PSR0900178410.hg.1 ENST00000373587.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0900178410.hg.1 ENST00000373588.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0900178410.hg.1 NM_004959.4 Refseq
          Affymetrix Human Clariom D Assay PSR0900178410.hg.1 TC0900011499.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0900178411.hg.1 ENST00000373587.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0900178411.hg.1 ENST00000373588.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0900178411.hg.1 ENST00000620110.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0900178411.hg.1 NM_004959.4 Refseq
          Affymetrix Human Clariom D Assay PSR0900178411.hg.1 ENST00000373587.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0900178411.hg.1 ENST00000373588.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0900178411.hg.1 ENST00000620110.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0900178411.hg.1 NM_004959.4 Refseq
          Affymetrix Human Clariom D Assay PSR0900178411.hg.1 TC0900011499.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0900011499.hg.1 ENST00000373587.3 Ensembl
          Affymetrix Human Clariom D Assay TC0900011499.hg.1 ENST00000373587.3 Ensembl
          Affymetrix Human Clariom D Assay TC0900011499.hg.1 ENST00000373587.3 Ensembl
          Affymetrix Human Clariom D Assay TC0900011499.hg.1 ENST00000373587.3 Ensembl
          Affymetrix Human Clariom D Assay TC0900011499.hg.1 ENST00000373587.3 Ensembl
          Affymetrix Human Clariom D Assay TC0900011499.hg.1 ENST00000373587.3 Ensembl
          Affymetrix Human Clariom D Assay TC0900011499.hg.1 ENST00000373588.8 Ensembl
          Affymetrix Human Clariom D Assay TC0900011499.hg.1 ENST00000373588.8 Ensembl
          Affymetrix Human Clariom D Assay TC0900011499.hg.1 ENST00000373588.8 Ensembl
          Affymetrix Human Clariom D Assay TC0900011499.hg.1 ENST00000373588.8 Ensembl
          Affymetrix Human Clariom D Assay TC0900011499.hg.1 ENST00000373588.8 Ensembl
          Affymetrix Human Clariom D Assay TC0900011499.hg.1 ENST00000373588.8 Ensembl
          Affymetrix Human Clariom D Assay TC0900011499.hg.1 ENST00000373588.8 Ensembl
          Affymetrix Human Clariom D Assay TC0900011499.hg.1 ENST00000373588.8 Ensembl
          Affymetrix Human Clariom D Assay TC0900011499.hg.1 ENST00000373588.8 Ensembl
          Affymetrix Human Clariom D Assay TC0900011499.hg.1 ENST00000373588.8 Ensembl
          Affymetrix Human Clariom D Assay TC0900011499.hg.1 ENST00000373588.8 Ensembl
          Affymetrix Human Clariom D Assay TC0900011499.hg.1 ENST00000455734.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900011499.hg.1 ENST00000455734.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900011499.hg.1 ENST00000455734.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900011499.hg.1 ENST00000455734.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900011499.hg.1 ENST00000620110.4 Ensembl
          Affymetrix Human Clariom D Assay TC0900011499.hg.1 ENST00000620110.4 Ensembl
          Affymetrix Human Clariom D Assay TC0900011499.hg.1 ENST00000620110.4 Ensembl
          Affymetrix Human Clariom D Assay TC0900011499.hg.1 ENST00000620110.4 Ensembl
          Affymetrix Human Clariom D Assay TC0900011499.hg.1 ENST00000620110.4 Ensembl
          Affymetrix Human Clariom D Assay TC0900011499.hg.1 ENST00000620110.4 Ensembl
          Affymetrix Human Clariom D Assay TC0900011499.hg.1 ENST00000620110.4 Ensembl
          Affymetrix Human Clariom D Assay TC0900011499.hg.1 ENST00000620110.4 Ensembl
          Affymetrix Human Clariom D Assay TC0900011499.hg.1 ENST00000620110.4 Ensembl
          Affymetrix Human Clariom D Assay TC0900011499.hg.1 ENST00000620110.4 Ensembl
          Affymetrix Human Clariom D Assay TC0900011499.hg.1 NM_004959.4 Refseq
          Affymetrix Human Clariom D Assay TC0900011499.hg.1 NM_004959.4 Refseq
          Affymetrix Human Clariom D Assay TC0900011499.hg.1 NM_004959.4 Refseq
          Affymetrix Human Clariom D Assay TC0900011499.hg.1 NM_004959.4 Refseq
          Affymetrix Human Clariom D Assay TC0900011499.hg.1 NM_004959.4 Refseq
          Affymetrix Human Clariom D Assay TC0900011499.hg.1 NM_004959.4 Refseq
          Affymetrix Human Clariom D Assay TC0900011499.hg.1 NM_004959.4 Refseq
          Affymetrix Human Clariom D Assay TC0900011499.hg.1 NM_004959.4 Refseq
          Affymetrix Human Clariom D Assay TC0900011499.hg.1 NM_004959.4 Refseq
          Affymetrix Human Clariom D Assay TC0900011499.hg.1 NM_004959.4 Refseq
          Affymetrix Human Clariom D Assay TC0900011499.hg.1 NM_004959.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline