Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01020682_m1
          See other QPCTL GE Assays ›
          Gene Symbol:
          QPCTL
          Gene Name:
          glutaminyl-peptide cyclotransferase like
          Gene Aliases:
          gQC
          Chromosome Location:
          Chr.19: 45692402 - 45703990 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01020682_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: QPCTL
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001163377.1 NP_001156849.1 2 - 3 566 68
          GenBank mRNA BC011553.1 - 2 - 3 352 4564051 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1900008343.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC19000653.hg.1

          Set Membership:

          Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons

          Panther Classification:

          Molecular Function -

          transferase

          Gene Ontology Categories:

          Function(s) Process(es)

          peptidyl-pyroglutamic acid biosynthetic process, using glutaminyl-peptide cyclotransferase
          zinc ion binding
          glutaminyl-peptide cyclotransferase activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1900008343.hg.1 ENST00000012049.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008343.hg.1 ENST00000012049.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008343.hg.1 ENST00000012049.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008343.hg.1 ENST00000012049.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008343.hg.1 ENST00000012049.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008343.hg.1 ENST00000012049.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008343.hg.1 ENST00000012049.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008343.hg.1 ENST00000012049.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008343.hg.1 ENST00000012049.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008343.hg.1 ENST00000012049.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008343.hg.1 ENST00000012049.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008343.hg.1 ENST00000012049.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008343.hg.1 ENST00000012049.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008343.hg.1 ENST00000012049.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008343.hg.1 ENST00000012049.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008343.hg.1 ENST00000366382.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008343.hg.1 ENST00000366382.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008343.hg.1 ENST00000366382.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008343.hg.1 ENST00000366382.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008343.hg.1 ENST00000366382.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008343.hg.1 ENST00000366382.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008343.hg.1 ENST00000366382.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008343.hg.1 ENST00000366382.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008343.hg.1 ENST00000366382.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008343.hg.1 ENST00000366382.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008343.hg.1 ENST00000366382.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008343.hg.1 ENST00000366382.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008343.hg.1 ENST00000591606.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008343.hg.1 ENST00000591606.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008343.hg.1 ENST00000591606.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008343.hg.1 ENST00000591606.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008343.hg.1 ENST00000591606.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008343.hg.1 ENST00000592769.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008343.hg.1 ENST00000592769.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008343.hg.1 ENST00000592769.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008343.hg.1 ENST00000592769.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008343.hg.1 ENST00000592769.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008343.hg.1 NM_001163377.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900008343.hg.1 NM_001163377.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900008343.hg.1 NM_001163377.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900008343.hg.1 NM_001163377.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900008343.hg.1 NM_001163377.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900008343.hg.1 NM_001163377.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900008343.hg.1 NM_001163377.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900008343.hg.1 NM_001163377.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900008343.hg.1 NM_001163377.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900008343.hg.1 NM_001163377.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900008343.hg.1 NM_001163377.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900008343.hg.1 NM_001163377.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900008343.hg.1 NM_001163377.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900008343.hg.1 NM_017659.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900008343.hg.1 NM_017659.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900008343.hg.1 NM_017659.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900008343.hg.1 NM_017659.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900008343.hg.1 NM_017659.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900008343.hg.1 NM_017659.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900008343.hg.1 NM_017659.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900008343.hg.1 NM_017659.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900008343.hg.1 NM_017659.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900008343.hg.1 NM_017659.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900008343.hg.1 NM_017659.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900008343.hg.1 NM_017659.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900008343.hg.1 NM_017659.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900008343.hg.1 NM_017659.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900008343.hg.1 NM_017659.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline