Search
Search
| Gene Symbol: FBN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001321431.2 | NP_001308360.1 | 55 - 56 | 7014 | 106 | |
| NM_032447.5 | NP_115823.3 | 55 - 56 | 7035 | 106 | ||
| XM_017027372.2 | XP_016882861.1 | 55 - 56 | 7008 | 106 | ||
| XM_017027373.2 | XP_016882862.1 | 55 - 56 | 6954 | 106 | ||
| XM_017027374.3 | XP_016882863.1 | 54 - 55 | 6918 | 106 | ||
| XM_017027375.3 | XP_016882864.1 | 54 - 55 | 6891 | 106 | ||
| XM_017027376.2 | XP_016882865.1 | 54 - 55 | 6885 | 106 | ||
| XM_017027377.3 | XP_016882866.1 | 36 - 37 | 4339 | 106 | ||
| XM_017027378.3 | XP_016882867.1 | 28 - 29 | 3481 | 106 | ||
| XM_017027379.2 | XP_016882868.1 | 56 - 57 | 7140 | 106 | ||
| GenBank mRNA | AB053450.2 | - | 54 - 55 | 6895 | 106 | |
| AK096539.1 | - | 10 - 11 | 1216 | 106 | ||
| AK131282.1 | - | 13 - 14 | 1556 | 106 | ||
| AY165865.1 | - | 54 - 55 | 7160 | 106 | ||
| BC153882.1 | - | 54 - 55 | 6895 | 106 | ||
Set Membership: |
|