| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07004535.hg.1 |
ENST00000275525.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07004535.hg.1 |
ENST00000457280.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07004535.hg.1 |
NM_000596.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07004535.hg.1 |
TC07000294.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007434.hg.1 |
ENST00000275525.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007434.hg.1 |
ENST00000275525.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007434.hg.1 |
ENST00000275525.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007434.hg.1 |
ENST00000275525.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007434.hg.1 |
ENST00000275525.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007434.hg.1 |
ENST00000275525.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007434.hg.1 |
ENST00000275525.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007434.hg.1 |
ENST00000275525.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007434.hg.1 |
ENST00000275525.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007434.hg.1 |
ENST00000457280.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007434.hg.1 |
ENST00000457280.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007434.hg.1 |
ENST00000457280.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007434.hg.1 |
ENST00000457280.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007434.hg.1 |
ENST00000457280.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007434.hg.1 |
ENST00000457280.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007434.hg.1 |
ENST00000457280.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007434.hg.1 |
ENST00000457280.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007434.hg.1 |
ENST00000468955.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007434.hg.1 |
ENST00000468955.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007434.hg.1 |
ENST00000468955.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007434.hg.1 |
ENST00000468955.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007434.hg.1 |
ENST00000468955.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007434.hg.1 |
ENST00000468955.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007434.hg.1 |
ENST00000468955.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007434.hg.1 |
NM_000596.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007434.hg.1 |
NM_000596.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007434.hg.1 |
NM_000596.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007434.hg.1 |
NM_000596.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007434.hg.1 |
NM_000596.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007434.hg.1 |
NM_000596.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007434.hg.1 |
NM_000596.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007434.hg.1 |
NM_000596.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007434.hg.1 |
NM_000596.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07004534.hg.1 |
ENST00000275525.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07004534.hg.1 |
ENST00000457280.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07004534.hg.1 |
NM_000596.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07004534.hg.1 |
TC07000294.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0700007434.hg.1 |
ENST00000275525.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007434.hg.1 |
ENST00000275525.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007434.hg.1 |
ENST00000275525.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007434.hg.1 |
ENST00000275525.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007434.hg.1 |
ENST00000275525.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007434.hg.1 |
ENST00000275525.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007434.hg.1 |
ENST00000275525.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007434.hg.1 |
ENST00000275525.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007434.hg.1 |
ENST00000275525.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007434.hg.1 |
ENST00000457280.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007434.hg.1 |
ENST00000457280.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007434.hg.1 |
ENST00000457280.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007434.hg.1 |
ENST00000457280.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007434.hg.1 |
ENST00000457280.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007434.hg.1 |
ENST00000457280.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007434.hg.1 |
ENST00000457280.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007434.hg.1 |
ENST00000457280.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007434.hg.1 |
ENST00000468955.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007434.hg.1 |
ENST00000468955.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007434.hg.1 |
ENST00000468955.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007434.hg.1 |
ENST00000468955.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007434.hg.1 |
ENST00000468955.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007434.hg.1 |
ENST00000468955.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007434.hg.1 |
ENST00000468955.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007434.hg.1 |
NM_000596.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700007434.hg.1 |
NM_000596.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700007434.hg.1 |
NM_000596.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700007434.hg.1 |
NM_000596.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700007434.hg.1 |
NM_000596.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700007434.hg.1 |
NM_000596.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700007434.hg.1 |
NM_000596.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700007434.hg.1 |
NM_000596.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700007434.hg.1 |
NM_000596.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700151780.hg.1 |
ENST00000275525.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700151780.hg.1 |
ENST00000457280.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700151780.hg.1 |
NM_000596.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700151780.hg.1 |
ENST00000275525.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700151780.hg.1 |
ENST00000457280.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700151780.hg.1 |
NM_000596.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700151780.hg.1 |
TC0700007434.hg.1 |
Clariom D TC |