Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01076320_g1
          See other LSR GE Assays ›
          Gene Symbol:
          LSR
          Gene Name:
          lipolysis stimulated lipoprotein receptor
          Gene Aliases:
          ILDR3, LISCH7
          Chromosome Location:
          Chr.19: 35248656 - 35267964 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01076320_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LSR
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001260490.1 NP_001247419.1 4 - 5 1062 90
          NM_015925.6 NP_057009.3 6 - 7 1329 90
          NM_205834.3 NP_991403.1 7 - 8 1386 90
          NM_205835.3 NP_991404.1 5 - 6 1182 90
          XM_011527026.1 XP_011525328.1 6 - 7 1214 90
          GenBank mRNA AK075426.1 - 6 - 7 981 90
          AK130367.1 - 5 - 6 836 90
          AK296618.1 - 4 - 5 716 90
          BC000015.2 - 6 - 7 960 90
          BC004381.1 - 6 - 7 977 90
          BC047376.1 - 7 - 8 1322 5272128 90

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1900007844.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC19000462.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene LSR

          Panther Classification:

          Molecular Function -

          defense/immunity protein immunoglobulin receptor superfamily

          Gene Ontology Categories:

          Function(s)

          liver development
          regulation of lipid metabolic process
          establishment of blood-brain barrier
          tricellular tight junction assembly

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR19006072.hg.1 ENST00000347609.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19006072.hg.1 ENST00000354900.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19006072.hg.1 ENST00000360798.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19006072.hg.1 ENST00000361790.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19006072.hg.1 ENST00000427250.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19006072.hg.1 NM_015925.6 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR19006072.hg.1 NM_205834.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR19006072.hg.1 NM_205835.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR19006072.hg.1 TC19000462.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1900157966.hg.1 ENST00000347609.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1900157966.hg.1 ENST00000354900.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1900157966.hg.1 ENST00000360798.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1900157966.hg.1 ENST00000361790.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1900157966.hg.1 ENST00000427250.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900157966.hg.1 ENST00000597446.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900157966.hg.1 ENST00000602122.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900157966.hg.1 ENST00000605618.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900157966.hg.1 ENST00000621372.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1900157966.hg.1 NM_001260489.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900157966.hg.1 NM_001260490.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900157966.hg.1 NM_015925.6 Refseq
          Affymetrix Human Clariom D Assay PSR1900157966.hg.1 NM_205834.3 Refseq
          Affymetrix Human Clariom D Assay PSR1900157966.hg.1 NM_205835.3 Refseq
          Affymetrix Human Clariom D Assay PSR1900157966.hg.1 TC1900007844.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1900157966.hg.1 ENST00000347609.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1900157966.hg.1 ENST00000354900.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1900157966.hg.1 ENST00000360798.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1900157966.hg.1 ENST00000361790.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1900157966.hg.1 ENST00000427250.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900157966.hg.1 ENST00000597446.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900157966.hg.1 ENST00000602122.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900157966.hg.1 ENST00000605618.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900157966.hg.1 ENST00000621372.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1900157966.hg.1 NM_001260489.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900157966.hg.1 NM_001260490.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900157966.hg.1 NM_015925.6 Refseq
          Affymetrix Human Clariom D Assay PSR1900157966.hg.1 NM_205834.3 Refseq
          Affymetrix Human Clariom D Assay PSR1900157966.hg.1 NM_205835.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline