Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01552701_g1
          See other LDB1 GE Assays ›
          Gene Symbol:
          LDB1
          Gene Name:
          LIM domain binding 1
          Gene Aliases:
          CLIM-2, CLIM2, LDB-1, NLI
          Chromosome Location:
          Chr.10: 102102100 - 102120473 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01552701_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LDB1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001113407.2 NP_001106878.1 2 - 3 487 63
          NM_001321612.1 NP_001308541.1 2 - 3 487 63
          NM_003893.4 NP_003884.1 2 - 3 660 63
          XM_017016867.1 XP_016872356.1 2 - 3 195 63
          XM_017016868.1 XP_016872357.1 2 - 3 195 63
          GenBank mRNA AB016485.1 - 1 - 2 66 63
          AF064491.1 - 2 - 3 329 63
          AF068652.1 - 2 - 3 300 63
          AK300588.1 - 2 - 3 527 63
          AK308481.1 - 2 - 3 314 63
          BC000482.2 - 2 - 3 639 2964724 63
          BC009246.2 - 2 - 3 639 2964724 63
          CB988590.1 - 2 - 3 514 63
          CV575131.1 - 2 - 3 467 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1000011669.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC10001617.hg.1

          Additional Information:

          This assay is in the 5'UTR region of the AB016485.1,AF064491.1,AF068652.1,AK300588.1,BC000482.2,BC009246.2,NM_003893.4 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene LDB1

          Panther Classification:

          Molecular Function -

          transcription cofactor

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription-dependent tethering of RNA polymerase II gene DNA at nuclear periphery
          gastrulation with mouth forming second
          hair follicle development
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          transcription from RNA polymerase II promoter
          multicellular organism development
          anterior/posterior axis specification
          epithelial structure maintenance
          Wnt signaling pathway
          cerebellar Purkinje cell differentiation
          cellular component assembly
          neuron differentiation
          regulation of cell migration
          regulation of DNA-templated transcription, elongation
          somatic stem cell population maintenance
          regulation of kinase activity
          histone H3-K4 acetylation
          negative regulation of erythrocyte differentiation
          positive regulation of cell adhesion
          negative regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of hemoglobin biosynthetic process
          regulation of focal adhesion assembly
          RNA polymerase II activating transcription factor binding
          enhancer sequence-specific DNA binding
          chromatin binding
          transcription corepressor activity
          protein binding
          enzyme binding
          LIM domain binding
          protein homodimerization activity
          protein self-association

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR10021052.hg.1 ENST00000361198.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10021052.hg.1 ENST00000425280.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10021052.hg.1 ENST00000490751.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10021052.hg.1 NM_001113407.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10021052.hg.1 NM_003893.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10021052.hg.1 TC10001617.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1000179877.hg.1 ENST00000361198.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1000179877.hg.1 ENST00000425280.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1000179877.hg.1 ENST00000490751.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1000179877.hg.1 NM_001113407.2 Refseq
          Affymetrix Human Clariom D Assay PSR1000179877.hg.1 NM_003893.4 Refseq
          Affymetrix Human Clariom D Assay PSR1000179877.hg.1 ENST00000361198.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1000179877.hg.1 ENST00000425280.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1000179877.hg.1 ENST00000490751.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1000179877.hg.1 NM_001113407.2 Refseq
          Affymetrix Human Clariom D Assay PSR1000179877.hg.1 NM_003893.4 Refseq
          Affymetrix Human Clariom D Assay PSR1000179877.hg.1 TC1000011669.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline