| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19002502.hg.1 |
ENST00000171214.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19002502.hg.1 |
NM_015725.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19002502.hg.1 |
TC19000166.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006964.hg.1 |
ENST00000587782.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006964.hg.1 |
ENST00000587782.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006964.hg.1 |
ENST00000587782.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006964.hg.1 |
ENST00000587782.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006964.hg.1 |
ENST00000589570.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006964.hg.1 |
ENST00000589570.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006964.hg.1 |
ENST00000589570.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006964.hg.1 |
ENST00000589570.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006964.hg.1 |
ENST00000591589.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006964.hg.1 |
ENST00000591589.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006964.hg.1 |
ENST00000591589.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006964.hg.1 |
ENST00000591589.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006964.hg.1 |
ENST00000591589.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006964.hg.1 |
ENST00000591589.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006964.hg.1 |
ENST00000591589.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006964.hg.1 |
ENST00000591589.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006964.hg.1 |
ENST00000591589.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006964.hg.1 |
ENST00000591589.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006964.hg.1 |
ENST00000591589.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006964.hg.1 |
ENST00000591589.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006964.hg.1 |
ENST00000591589.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006964.hg.1 |
NM_015725.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006964.hg.1 |
NM_015725.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006964.hg.1 |
NM_015725.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006964.hg.1 |
NM_015725.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006964.hg.1 |
NM_015725.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006964.hg.1 |
NM_015725.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006964.hg.1 |
NM_015725.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006964.hg.1 |
NM_015725.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006964.hg.1 |
NM_015725.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006964.hg.1 |
NM_015725.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006964.hg.1 |
NM_015725.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006964.hg.1 |
NM_015725.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006964.hg.1 |
NM_015725.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900149361.hg.1 |
ENST00000591589.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900149361.hg.1 |
NM_015725.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900149361.hg.1 |
TC1900006964.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900149361.hg.1 |
ENST00000591589.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900149361.hg.1 |
NM_015725.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900006964.hg.1 |
ENST00000587782.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006964.hg.1 |
ENST00000587782.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006964.hg.1 |
ENST00000587782.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006964.hg.1 |
ENST00000587782.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006964.hg.1 |
ENST00000589570.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006964.hg.1 |
ENST00000589570.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006964.hg.1 |
ENST00000589570.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006964.hg.1 |
ENST00000589570.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006964.hg.1 |
ENST00000591589.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006964.hg.1 |
ENST00000591589.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006964.hg.1 |
ENST00000591589.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006964.hg.1 |
ENST00000591589.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006964.hg.1 |
ENST00000591589.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006964.hg.1 |
ENST00000591589.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006964.hg.1 |
ENST00000591589.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006964.hg.1 |
ENST00000591589.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006964.hg.1 |
ENST00000591589.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006964.hg.1 |
ENST00000591589.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006964.hg.1 |
ENST00000591589.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006964.hg.1 |
ENST00000591589.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006964.hg.1 |
ENST00000591589.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006964.hg.1 |
NM_015725.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900006964.hg.1 |
NM_015725.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900006964.hg.1 |
NM_015725.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900006964.hg.1 |
NM_015725.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900006964.hg.1 |
NM_015725.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900006964.hg.1 |
NM_015725.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900006964.hg.1 |
NM_015725.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900006964.hg.1 |
NM_015725.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900006964.hg.1 |
NM_015725.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900006964.hg.1 |
NM_015725.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900006964.hg.1 |
NM_015725.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900006964.hg.1 |
NM_015725.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900006964.hg.1 |
NM_015725.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900149360.hg.1 |
ENST00000589570.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900149360.hg.1 |
ENST00000591589.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900149360.hg.1 |
NM_015725.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900149360.hg.1 |
TC1900006964.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900149360.hg.1 |
ENST00000589570.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900149360.hg.1 |
ENST00000591589.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900149360.hg.1 |
NM_015725.2 |
Refseq |