Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01565273_m1
          See other FN1 GE Assays ›
          Gene Symbol:
          FN1
          Gene Name:
          fibronectin 1
          Gene Aliases:
          CIG, ED-B, FINC, FN, FNZ, GFND, GFND2, LETS, MSF, SMDCF
          Chromosome Location:
          Chr.2: 215360865 - 215436068 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01565273_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: FN1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001306130.2 NP_001293059.2 38 - 39 6242 92
          NM_001365524.2 NP_001352453.1 38 - 39 6239 92
          NM_212474.3 NP_997639.2 37 - 38 5969 92
          GenBank mRNA AF130095.1 - 3 - 4 535 92
          AJ320525.1 - 5 - 6 612 92
          AJ320527.1 - 5 - 6 612 92
          AK300458.1 - 17 - 18 2590 92
          AL832202.1 - 37 - 38 5970 92
          AL832771.1 - 37 - 38 5970 92
          BC117176.1 - 37 - 38 5949 92
          BC143763.1 - 37 - 38 5949 92
          CR749316.1 - 38 - 39 6241 92
          EF550133.1 - 9 - 10 1338 92
          GQ891409.1 - 37 - 38 5949 92

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0200015650.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC02002747.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene FN1

          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          regulation of protein phosphorylation
          acute-phase response
          cell-substrate junction assembly
          cell adhesion
          cell-matrix adhesion
          calcium-independent cell-matrix adhesion
          integrin-mediated signaling pathway
          nervous system development
          heart development
          positive regulation of cell proliferation
          regulation of cell shape
          response to wounding
          positive regulation of gene expression
          response to muscle activity
          integrin activation
          substrate adhesion-dependent cell spreading
          endodermal cell differentiation
          wound healing
          endothelial cell migration
          positive regulation of axon extension
          positive regulation of fibroblast proliferation
          interaction with symbiont
          positive regulation of protein kinase B signaling
          regulation of ERK1 and ERK2 cascade
          negative regulation of transforming growth factor beta production
          blood coagulation, fibrin clot formation
          negative regulation of monocyte activation
          neural crest cell migration involved in autonomic nervous system development
          positive regulation of substrate-dependent cell migration, cell attachment to substrate
          protease binding
          receptor binding
          integrin binding
          extracellular matrix structural constituent
          protein binding
          collagen binding
          heparin binding
          peptidase activator activity
          identical protein binding
          proteoglycan binding
          receptor agonist activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR02044130.hg.1 ENST00000265313.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02044130.hg.1 ENST00000323926.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02044130.hg.1 ENST00000336916.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02044130.hg.1 ENST00000346544.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02044130.hg.1 ENST00000354785.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02044130.hg.1 ENST00000356005.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02044130.hg.1 ENST00000357009.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02044130.hg.1 ENST00000357867.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02044130.hg.1 ENST00000359671.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02044130.hg.1 ENST00000421182.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02044130.hg.1 ENST00000432072.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02044130.hg.1 ENST00000443816.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02044130.hg.1 ENST00000446046.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02044130.hg.1 NM_002026.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02044130.hg.1 NM_212474.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02044130.hg.1 NM_212476.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02044130.hg.1 NM_212478.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02044130.hg.1 NM_212482.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02044130.hg.1 TC02002747.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0200214827.hg.1 ENST00000323926.10 Ensembl
          Affymetrix Human Clariom D Assay PSR0200214827.hg.1 ENST00000336916.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0200214827.hg.1 ENST00000354785.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0200214827.hg.1 ENST00000356005.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0200214827.hg.1 ENST00000357867.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0200214827.hg.1 ENST00000359671.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0200214827.hg.1 ENST00000421182.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0200214827.hg.1 ENST00000432072.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0200214827.hg.1 ENST00000438981.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200214827.hg.1 ENST00000443816.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0200214827.hg.1 ENST00000446046.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0200214827.hg.1 ENST00000456923.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0200214827.hg.1 ENST00000460217.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200214827.hg.1 ENST00000461974.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200214827.hg.1 ENST00000485567.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200214827.hg.1 ENST00000492816.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0200214827.hg.1 NM_001306129.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200214827.hg.1 NM_001306130.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200214827.hg.1 NM_001306131.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200214827.hg.1 NM_001306132.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200214827.hg.1 NM_002026.3 Refseq
          Affymetrix Human Clariom D Assay PSR0200214827.hg.1 NM_212474.2 Refseq
          Affymetrix Human Clariom D Assay PSR0200214827.hg.1 NM_212476.2 Refseq
          Affymetrix Human Clariom D Assay PSR0200214827.hg.1 NM_212478.2 Refseq
          Affymetrix Human Clariom D Assay PSR0200214827.hg.1 NM_212482.2 Refseq
          Affymetrix Human Clariom D Assay PSR0200214827.hg.1 ENST00000323926.10 Ensembl
          Affymetrix Human Clariom D Assay PSR0200214827.hg.1 ENST00000336916.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0200214827.hg.1 ENST00000354785.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0200214827.hg.1 ENST00000356005.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0200214827.hg.1 ENST00000357867.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0200214827.hg.1 ENST00000359671.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0200214827.hg.1 ENST00000421182.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0200214827.hg.1 ENST00000432072.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0200214827.hg.1 ENST00000438981.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200214827.hg.1 ENST00000443816.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0200214827.hg.1 ENST00000446046.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0200214827.hg.1 ENST00000456923.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0200214827.hg.1 ENST00000460217.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200214827.hg.1 ENST00000461974.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200214827.hg.1 ENST00000485567.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200214827.hg.1 ENST00000492816.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0200214827.hg.1 NM_001306129.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200214827.hg.1 NM_001306130.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200214827.hg.1 NM_001306131.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200214827.hg.1 NM_001306132.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200214827.hg.1 NM_002026.3 Refseq
          Affymetrix Human Clariom D Assay PSR0200214827.hg.1 NM_212474.2 Refseq
          Affymetrix Human Clariom D Assay PSR0200214827.hg.1 NM_212476.2 Refseq
          Affymetrix Human Clariom D Assay PSR0200214827.hg.1 NM_212478.2 Refseq
          Affymetrix Human Clariom D Assay PSR0200214827.hg.1 NM_212482.2 Refseq
          Affymetrix Human Clariom D Assay PSR0200214827.hg.1 TC0200015650.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline