Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01589322_s1
          See other PEX39 GE Assays ›
          Gene Symbol:
          PEX39
          Gene Name:
          peroxisomal biogenesis factor 39
          Gene Aliases:
          C6orf226
          Chromosome Location:
          Chr.6: 42890265 - 42890821 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01589322_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PEX39
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001008739.2 NP_001008739.1 1 436 101
          GenBank mRNA BC051007.1 - 1 431 101
          BC060325.1 - 1 419 101
          BC073804.1 - 1 431 101
          BC094854.1 - 1 431 101

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0600011846.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC06001738.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene PEX39

          Gene Ontology Categories:

          Process(es)

          protein binding

          MicroArray Probes:

          Agilent Affymetrix ArrayStar Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04694025 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04694026 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24888923 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04694034 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04694036 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24888925 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24888933 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04694030 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24888922 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04694033 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24888929 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24888932 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 523160 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3408898 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_32_P149174 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_32_P149174 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 13212 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 40860 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 2963 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P01467514 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P01467514 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3408898 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_32_P149174 - -
          SurePrint G3 Human GE Microarray G4851A A_32_P149174 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3408898 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04694037 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24888932 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04694036 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04694025 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24888925 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24888937 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04694026 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04694034 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24888923 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04694033 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24888937 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04694033 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04694037 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04694036 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04694034 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04694026 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04694025 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24888923 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24888925 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24888932 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P04694029 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P04694036 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P04694025 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P04694033 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P04694034 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P04694031 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P04694037 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P15810466 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P04694033 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P15810464 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P04694034 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P04694031 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P15810465 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P04694030 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P04694032 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P04694035 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_32_P149174 - -
          Agilent Human 1B Microarray G4111A A_32_P149174 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04694009 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04694011 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04694013 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04694034 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04694029 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15810466 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04694036 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04694031 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04694006 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04694033 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04694004 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04694026 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04694008 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array NM_001008739_at - -
          Affymetrix Human Gene 1.0 ST Array 8126449 - -
          Affymetrix Human Gene 1.0 ST Array 8126448 - -
          Affymetrix Human Gene 1.0 ST Array 8126447 - -
          Affymetrix Human Gene 1.1 ST Array 8126446 - -
          Affymetrix HT HG-U133+ PM Array Plate 228458_PM_at - -
          Affymetrix Human Genome U219 Array 11724332_PM_at - -
          Affymetrix Human Genome U219 Array 11724332_at - -
          Affymetrix Human Transcriptome Array 2.0 TC0600011846.hg.1 ENST00000408925.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600011846.hg.1 ENST00000408925.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600011846.hg.1 NM_001008739.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600011846.hg.1 NM_001008739.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06020745.hg.1 ENST00000408925.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06020745.hg.1 NM_001008739.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06020745.hg.1 TC06001738.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0600181295.hg.1 ENST00000408925.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0600181295.hg.1 NM_001008739.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600181295.hg.1 ENST00000408925.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0600181295.hg.1 NM_001008739.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600181295.hg.1 TC0600011846.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0600011846.hg.1 ENST00000408925.3 Ensembl
          Affymetrix Human Clariom D Assay TC0600011846.hg.1 ENST00000408925.3 Ensembl
          Affymetrix Human Clariom D Assay TC0600011846.hg.1 NM_001008739.1 Refseq
          Affymetrix Human Clariom D Assay TC0600011846.hg.1 NM_001008739.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600181296.hg.1 ENST00000408925.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0600181296.hg.1 NM_001008739.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600181296.hg.1 ENST00000408925.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0600181296.hg.1 NM_001008739.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600181296.hg.1 TC0600011846.hg.1 Clariom D TC
          Human Genome U133 Plus 2.0 Array 228458_PM_at - -
          Human Genome U133 Plus 2.0 Array 228458_at - -
          Human Genome U133 Plus 2.0 Array 1561770_PM_at - -
          Human Genome U133 Plus 2.0 Array 1561770_at - -
          Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) NM_001008739_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 228458_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 228458_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 228458_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 228458_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 228458_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 228458_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 228458_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 228458_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 228458_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 228458_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 228458_at2 - -
          Human Genome U95 Set 90236_at - -
          Human Genome U95 Set 90236_PM_at - -
          Human Genome U133 Set 228458_PM_at - -
          Human Genome U133 Set 228458_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A029917 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A029917 - -
          Array Name Probe IDs Targets Target Type
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1743755 BC073804.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1743755 BC094854.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1743755 NM_001008739.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1743755 BC051007.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1743755 BC060325.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1743755 BC073804.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1743755 BC094854.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1743755 NM_001008739.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1743755 BC051007.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1743755 BC060325.1 human_transcriptome
          Illumina HumanRef-8 v2 Expression BeadChip GI_15465-S - -
          Illumina human-6 v2.0 expression beadchip ILMN_1743755 BC073804.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1743755 BC094854.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1743755 NM_001008739.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1743755 BC051007.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1743755 BC060325.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1743755 BC073804.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1743755 BC094854.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1743755 NM_001008739.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1743755 BC051007.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1743755 BC060325.1 human_transcriptome
          Illumina Sentrix Human-6 v2 Expression BeadChip 4010605 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_15465 - -
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1743755 BC073804.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1743755 BC094854.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1743755 NM_001008739.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1743755 BC051007.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1743755 BC060325.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1743755 BC073804.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1743755 BC094854.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1743755 NM_001008739.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1743755 BC051007.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1743755 BC060325.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1743755 BC073804.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1743755 BC094854.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1743755 NM_001008739.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1743755 BC051007.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1743755 BC060325.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1743755 BC073804.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1743755 BC094854.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1743755 NM_001008739.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1743755 BC051007.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1743755 BC060325.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline