Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01943223_g1
          See other OR2M5 GE Assays ›
          Gene Symbol:
          OR2M5
          Gene Name:
          olfactory receptor family 2 subfamily M member 5
          Gene Aliases:
          OR2M5P
          Chromosome Location:
          Chr.1: 248145148 - 248146086 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01943223_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: OR2M5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001004690.1 NP_001004690.1 1 879 162

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100012348.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC01002030.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Homo sapiens 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene OR2M5

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          G-protein coupled receptor activity
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15205342 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21005841 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21005874 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21005852 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15205329 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21005846 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00931540 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21005849 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00931532 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00931490 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21005825 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21005866 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15205311 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21005865 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21005885 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21005818 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21005851 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00931511 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21005869 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00931495 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21005834 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00931533 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21005830 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00931491 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21005878 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21005870 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15205372 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21005832 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00931517 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00931506 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21005800 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00931520 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21005880 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21005845 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21005843 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21005871 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21005861 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21005886 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21005840 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00931493 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21005828 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21005829 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00931536 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21005821 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21005820 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00931502 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00931523 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21005799 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15205328 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21005883 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00931494 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21005848 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00931504 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21005862 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21005827 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21005875 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21005826 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00931512 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3290249 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 53572 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3331952 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3290249 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3290249 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21005812 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00931495 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21005819 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00931506 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00931511 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21005846 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21005825 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00931517 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21005822 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21005800 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21005829 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21005830 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15205342 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21005851 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21005841 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00931494 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21005847 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21005808 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21005861 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21005845 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21005820 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21005843 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21005840 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21005828 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15205311 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00931491 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21005852 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21005799 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00931493 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00931512 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21005834 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21005818 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15205347 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15205329 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21005849 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00931520 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21005832 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00931504 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21005799 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21005845 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21005861 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21005852 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00931495 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21005851 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00931517 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00931520 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21005834 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21005832 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00931494 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21005800 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00931506 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21005841 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00931504 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21005840 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00931493 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15205347 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15205342 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15205311 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21005812 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21005849 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21005846 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21005829 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21005825 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21005822 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00931511 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21005828 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21005847 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21005819 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21005820 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21005843 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00931491 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15205329 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21005830 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21005818 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21005808 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00931512 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00931504 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15205309 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00931495 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15205367 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00931521 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00931535 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15205371 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15205365 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00931497 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00931509 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00931494 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00931511 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15205364 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00931518 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00931501 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00931527 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00931540 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00931513 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00931490 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00931531 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00931537 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00931499 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15205368 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15205369 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00931512 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00931488 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00931505 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array NM_001004690_at - -
          Affymetrix Human Gene 1.0 ST Array 7911264 - -
          Affymetrix Human Gene 1.0 ST Array 7911262 - -
          Affymetrix Human Gene 1.0 ST Array 7911261 - -
          Affymetrix Human Gene 1.1 ST Array 7911260 - -
          Affymetrix Human Gene 1.1 ST Array 7911263 - -
          Affymetrix Human Genome U219 Array 11742592_PM_x_at - -
          Affymetrix Human Genome U219 Array 11742592_x_at - -
          Affymetrix Human Genome U219 Array 11742590_at - -
          Affymetrix Human Genome U219 Array 11742590_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR01031120.hg.1 ENST00000366476.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01031120.hg.1 NM_001004690.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01031120.hg.1 TC01002030.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0100190620.hg.1 ENST00000366476.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100190620.hg.1 NM_001004690.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100190620.hg.1 ENST00000366476.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100190620.hg.1 NM_001004690.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100190620.hg.1 TC0100012348.hg.1 Clariom D TC
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A042320 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A042320 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 148980 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1699872 NM_001004690.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1699872 NM_001004690.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1699872 NM_001004690.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1699872 NM_001004690.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1699872 NM_001004690.1 human_transcriptome
          Illumina HumanRef-8 v2 Expression BeadChip GI_23603-S - -
          Illumina human-6 v2.0 expression beadchip ILMN_1699872 NM_001004690.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1699872 NM_001004690.1 human_transcriptome
          Illumina Sentrix Human-6 v2 Expression BeadChip 3780092 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_23603 - -
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1699872 NM_001004690.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1699872 NM_001004690.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1699872 NM_001004690.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1699872 NM_001004690.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1699872 NM_001004690.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline