| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010525.hg.1 |
ENST00000295854.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010525.hg.1 |
ENST00000295854.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010525.hg.1 |
ENST00000295854.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010525.hg.1 |
ENST00000295854.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010525.hg.1 |
ENST00000302823.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010525.hg.1 |
ENST00000302823.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010525.hg.1 |
ENST00000302823.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010525.hg.1 |
ENST00000302823.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010525.hg.1 |
ENST00000302823.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010525.hg.1 |
ENST00000302823.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010525.hg.1 |
ENST00000302823.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010525.hg.1 |
ENST00000302823.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010525.hg.1 |
ENST00000302823.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010525.hg.1 |
ENST00000302823.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010525.hg.1 |
ENST00000302823.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010525.hg.1 |
ENST00000302823.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010525.hg.1 |
ENST00000302823.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010525.hg.1 |
ENST00000427473.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010525.hg.1 |
ENST00000427473.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010525.hg.1 |
ENST00000427473.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010525.hg.1 |
ENST00000427473.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010525.hg.1 |
ENST00000427473.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010525.hg.1 |
ENST00000427473.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010525.hg.1 |
ENST00000427473.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010525.hg.1 |
ENST00000427473.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010525.hg.1 |
ENST00000472206.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010525.hg.1 |
ENST00000472206.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010525.hg.1 |
ENST00000472206.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010525.hg.1 |
ENST00000472206.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010525.hg.1 |
ENST00000487393.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010525.hg.1 |
ENST00000487393.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010525.hg.1 |
ENST00000487393.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010525.hg.1 |
NM_001037631.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010525.hg.1 |
NM_001037631.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010525.hg.1 |
NM_001037631.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010525.hg.1 |
NM_001037631.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010525.hg.1 |
NM_001037631.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010525.hg.1 |
NM_001037631.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010525.hg.1 |
NM_001037631.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010525.hg.1 |
NM_001037631.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010525.hg.1 |
NM_001037631.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010525.hg.1 |
NM_001037631.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010525.hg.1 |
NM_001037631.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010525.hg.1 |
NM_001037631.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010525.hg.1 |
NM_005214.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010525.hg.1 |
NM_005214.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010525.hg.1 |
NM_005214.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010525.hg.1 |
NM_005214.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010525.hg.1 |
NM_005214.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010525.hg.1 |
NM_005214.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010525.hg.1 |
NM_005214.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010525.hg.1 |
NM_005214.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010525.hg.1 |
NM_005214.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010525.hg.1 |
NM_005214.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010525.hg.1 |
NM_005214.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010525.hg.1 |
NM_005214.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010525.hg.1 |
NM_005214.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02018471.hg.1 |
ENST00000295854.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02018471.hg.1 |
ENST00000302823.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02018471.hg.1 |
ENST00000427473.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02018471.hg.1 |
ENST00000472206.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02018471.hg.1 |
NM_001037631.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02018471.hg.1 |
NM_005214.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02018471.hg.1 |
TC02001201.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02018472.hg.1 |
ENST00000295854.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02018472.hg.1 |
ENST00000302823.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02018472.hg.1 |
ENST00000427473.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02018472.hg.1 |
NM_001037631.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02018472.hg.1 |
NM_005214.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02018472.hg.1 |
TC02001201.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0200010525.hg.1 |
ENST00000295854.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010525.hg.1 |
ENST00000295854.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010525.hg.1 |
ENST00000295854.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010525.hg.1 |
ENST00000295854.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010525.hg.1 |
ENST00000302823.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010525.hg.1 |
ENST00000302823.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010525.hg.1 |
ENST00000302823.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010525.hg.1 |
ENST00000302823.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010525.hg.1 |
ENST00000302823.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010525.hg.1 |
ENST00000302823.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010525.hg.1 |
ENST00000302823.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010525.hg.1 |
ENST00000302823.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010525.hg.1 |
ENST00000302823.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010525.hg.1 |
ENST00000302823.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010525.hg.1 |
ENST00000302823.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010525.hg.1 |
ENST00000302823.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010525.hg.1 |
ENST00000302823.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010525.hg.1 |
ENST00000427473.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010525.hg.1 |
ENST00000427473.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010525.hg.1 |
ENST00000427473.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010525.hg.1 |
ENST00000427473.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010525.hg.1 |
ENST00000427473.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010525.hg.1 |
ENST00000427473.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010525.hg.1 |
ENST00000427473.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010525.hg.1 |
ENST00000427473.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010525.hg.1 |
ENST00000472206.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010525.hg.1 |
ENST00000472206.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010525.hg.1 |
ENST00000472206.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010525.hg.1 |
ENST00000472206.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010525.hg.1 |
ENST00000487393.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010525.hg.1 |
ENST00000487393.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010525.hg.1 |
ENST00000487393.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010525.hg.1 |
NM_001037631.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200010525.hg.1 |
NM_001037631.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200010525.hg.1 |
NM_001037631.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200010525.hg.1 |
NM_001037631.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200010525.hg.1 |
NM_001037631.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200010525.hg.1 |
NM_001037631.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200010525.hg.1 |
NM_001037631.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200010525.hg.1 |
NM_001037631.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200010525.hg.1 |
NM_001037631.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200010525.hg.1 |
NM_001037631.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200010525.hg.1 |
NM_001037631.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200010525.hg.1 |
NM_001037631.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200010525.hg.1 |
NM_005214.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200010525.hg.1 |
NM_005214.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200010525.hg.1 |
NM_005214.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200010525.hg.1 |
NM_005214.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200010525.hg.1 |
NM_005214.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200010525.hg.1 |
NM_005214.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200010525.hg.1 |
NM_005214.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200010525.hg.1 |
NM_005214.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200010525.hg.1 |
NM_005214.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200010525.hg.1 |
NM_005214.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200010525.hg.1 |
NM_005214.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200010525.hg.1 |
NM_005214.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200010525.hg.1 |
NM_005214.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200174593.hg.1 |
ENST00000295854.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200174593.hg.1 |
ENST00000302823.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200174593.hg.1 |
ENST00000427473.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200174593.hg.1 |
NM_001037631.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200174593.hg.1 |
NM_005214.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200174593.hg.1 |
ENST00000295854.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200174593.hg.1 |
ENST00000302823.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200174593.hg.1 |
ENST00000427473.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200174593.hg.1 |
NM_001037631.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200174593.hg.1 |
NM_005214.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200174593.hg.1 |
TC0200010525.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0200174592.hg.1 |
ENST00000295854.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200174592.hg.1 |
ENST00000302823.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200174592.hg.1 |
ENST00000427473.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200174592.hg.1 |
ENST00000472206.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200174592.hg.1 |
NM_001037631.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200174592.hg.1 |
NM_005214.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200174592.hg.1 |
ENST00000295854.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200174592.hg.1 |
ENST00000302823.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200174592.hg.1 |
ENST00000427473.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200174592.hg.1 |
ENST00000472206.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200174592.hg.1 |
NM_001037631.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200174592.hg.1 |
NM_005214.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200174592.hg.1 |
TC0200010525.hg.1 |
Clariom D TC |