Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03655841_m1
          See other PCOLCE-AS1 GE Assays ›
          Gene Symbol:
          PCOLCE-AS1
          Gene Name:
          PCOLCE antisense RNA 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 100589401 - 100604038 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03655841_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price 434.00
          Your Price
          Online offer: 407.65
          407.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PCOLCE-AS1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_038910.1 6 - 7 1506 71
          GenBank mRNA AK097705.1 - 6 - 7 1506 71

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream F-box protein 24 FBXO24 FBX24 Anti-sense Chr. 7 - 100582490 - 100601117 on NCBI Build GRCh38
          Downstream procollagen C-endopeptidase enhancer PCOLCE PCPE,
          PCPE-1,
          PCPE1
          2.5kb Anti-sense Chr. 7 - 100602259 - 100608175 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0700012009.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC07001673.hg.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Homo sapiens Probe spans exons

          MicroArray Probes:

          Affymetrix ArrayStar

          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 1560939_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR07025780.hg.1 ENST00000442166.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07025780.hg.1 NR_038910.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR07025780.hg.1 TC07001673.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR07025781.hg.1 ENST00000442166.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07025781.hg.1 NR_038910.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR07025781.hg.1 TC07001673.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR07025782.hg.1 ENST00000442166.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07025782.hg.1 ENST00000544873.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07025782.hg.1 NR_038910.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR07025782.hg.1 TC07001673.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR07025779.hg.1 ENST00000442166.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07025779.hg.1 NR_038910.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR07025779.hg.1 TC07001673.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0700012009.hg.1 ENST00000442166.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700012009.hg.1 ENST00000442166.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700012009.hg.1 ENST00000442166.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700012009.hg.1 ENST00000442166.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700012009.hg.1 ENST00000442166.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700012009.hg.1 ENST00000442166.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700012009.hg.1 ENST00000442166.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700012009.hg.1 ENST00000442166.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700012009.hg.1 ENST00000442166.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700012009.hg.1 ENST00000442166.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700012009.hg.1 ENST00000442166.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700012009.hg.1 ENST00000442166.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700012009.hg.1 ENST00000442166.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700012009.hg.1 ENST00000442166.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700012009.hg.1 ENST00000446022.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700012009.hg.1 ENST00000446022.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700012009.hg.1 ENST00000446022.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700012009.hg.1 ENST00000446022.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700012009.hg.1 ENST00000544873.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700012009.hg.1 ENST00000544873.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700012009.hg.1 ENST00000544873.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700012009.hg.1 ENST00000544873.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700012009.hg.1 ENST00000544873.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700012009.hg.1 ENST00000544873.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700012009.hg.1 ENST00000544873.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700012009.hg.1 ENST00000544873.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700012009.hg.1 ENST00000544873.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700012009.hg.1 ENST00000544873.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700012009.hg.1 NR_038910.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700012009.hg.1 NR_038910.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700012009.hg.1 NR_038910.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700012009.hg.1 NR_038910.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700012009.hg.1 NR_038910.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700012009.hg.1 NR_038910.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700012009.hg.1 NR_038910.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700012009.hg.1 NR_038910.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700012009.hg.1 NR_038910.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700012009.hg.1 NR_038910.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700012009.hg.1 NR_038910.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700012009.hg.1 NR_038910.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700012009.hg.1 NR_038910.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700012009.hg.1 NR_038910.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700186220.hg.1 ENST00000442166.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0700186220.hg.1 NR_038910.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700186220.hg.1 ENST00000442166.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0700186220.hg.1 NR_038910.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700186220.hg.1 TC0700012009.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0700186219.hg.1 ENST00000442166.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0700186219.hg.1 NR_038910.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700186219.hg.1 ENST00000442166.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0700186219.hg.1 NR_038910.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700186219.hg.1 TC0700012009.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0700186221.hg.1 ENST00000442166.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0700186221.hg.1 ENST00000544873.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700186221.hg.1 NR_038910.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700186221.hg.1 ENST00000442166.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0700186221.hg.1 ENST00000544873.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700186221.hg.1 NR_038910.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700186221.hg.1 TC0700012009.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0700012009.hg.1 ENST00000442166.2 Ensembl
          Affymetrix Human Clariom D Assay TC0700012009.hg.1 ENST00000442166.2 Ensembl
          Affymetrix Human Clariom D Assay TC0700012009.hg.1 ENST00000442166.2 Ensembl
          Affymetrix Human Clariom D Assay TC0700012009.hg.1 ENST00000442166.2 Ensembl
          Affymetrix Human Clariom D Assay TC0700012009.hg.1 ENST00000442166.2 Ensembl
          Affymetrix Human Clariom D Assay TC0700012009.hg.1 ENST00000442166.2 Ensembl
          Affymetrix Human Clariom D Assay TC0700012009.hg.1 ENST00000442166.2 Ensembl
          Affymetrix Human Clariom D Assay TC0700012009.hg.1 ENST00000442166.2 Ensembl
          Affymetrix Human Clariom D Assay TC0700012009.hg.1 ENST00000442166.2 Ensembl
          Affymetrix Human Clariom D Assay TC0700012009.hg.1 ENST00000442166.2 Ensembl
          Affymetrix Human Clariom D Assay TC0700012009.hg.1 ENST00000442166.2 Ensembl
          Affymetrix Human Clariom D Assay TC0700012009.hg.1 ENST00000442166.2 Ensembl
          Affymetrix Human Clariom D Assay TC0700012009.hg.1 ENST00000442166.2 Ensembl
          Affymetrix Human Clariom D Assay TC0700012009.hg.1 ENST00000442166.2 Ensembl
          Affymetrix Human Clariom D Assay TC0700012009.hg.1 ENST00000446022.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700012009.hg.1 ENST00000446022.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700012009.hg.1 ENST00000446022.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700012009.hg.1 ENST00000446022.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700012009.hg.1 ENST00000544873.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700012009.hg.1 ENST00000544873.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700012009.hg.1 ENST00000544873.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700012009.hg.1 ENST00000544873.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700012009.hg.1 ENST00000544873.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700012009.hg.1 ENST00000544873.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700012009.hg.1 ENST00000544873.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700012009.hg.1 ENST00000544873.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700012009.hg.1 ENST00000544873.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700012009.hg.1 ENST00000544873.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700012009.hg.1 NR_038910.1 Refseq
          Affymetrix Human Clariom D Assay TC0700012009.hg.1 NR_038910.1 Refseq
          Affymetrix Human Clariom D Assay TC0700012009.hg.1 NR_038910.1 Refseq
          Affymetrix Human Clariom D Assay TC0700012009.hg.1 NR_038910.1 Refseq
          Affymetrix Human Clariom D Assay TC0700012009.hg.1 NR_038910.1 Refseq
          Affymetrix Human Clariom D Assay TC0700012009.hg.1 NR_038910.1 Refseq
          Affymetrix Human Clariom D Assay TC0700012009.hg.1 NR_038910.1 Refseq
          Affymetrix Human Clariom D Assay TC0700012009.hg.1 NR_038910.1 Refseq
          Affymetrix Human Clariom D Assay TC0700012009.hg.1 NR_038910.1 Refseq
          Affymetrix Human Clariom D Assay TC0700012009.hg.1 NR_038910.1 Refseq
          Affymetrix Human Clariom D Assay TC0700012009.hg.1 NR_038910.1 Refseq
          Affymetrix Human Clariom D Assay TC0700012009.hg.1 NR_038910.1 Refseq
          Affymetrix Human Clariom D Assay TC0700012009.hg.1 NR_038910.1 Refseq
          Affymetrix Human Clariom D Assay TC0700012009.hg.1 NR_038910.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700186218.hg.1 ENST00000442166.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0700186218.hg.1 NR_038910.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700186218.hg.1 ENST00000442166.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0700186218.hg.1 NR_038910.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700186218.hg.1 TC0700012009.hg.1 Clariom D TC
          Human Genome U133 Plus 2.0 Array 1560939_at - -
          Human Genome U133 Plus 2.0 Array 1560939_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560939_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560939_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560939_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560939_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560939_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560939_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560939_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560939_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560939_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560939_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560939_at1 - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A033261 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A033261 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-fvx7n:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline