Search
Search

MIR142HG| Gene Symbol: MIR142HG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_186557.1 | 1 | 368 | 89 | ||
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 142 | MIR142 |
MIRN142, mir-142 |
-611.0 | Sense | Chr. 17 - 58331232 - 58331318 on NCBI Build GRCh38 |
| Downstream | microRNA 4736 | MIR4736 | 3.5kb | Sense | Chr. 17 - 58335976 - 58336022 on NCBI Build GRCh38 |
Set Membership: |
|