Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04334488_m1
          See other ACBD5 GE Assays ›
          Gene Symbol:
          ACBD5
          Gene Name:
          acyl-CoA binding domain containing 5
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 27182624 - 27248674 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04334488_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ACBD5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001042473.3 NP_001035938.1 12 - 13 1631 107
          NM_001271512.3 NP_001258441.1 11 - 12 1945 107
          NM_001301251.1 NP_001288180.1 11 - 12 1510 107
          NM_001301252.1 NP_001288181.1 10 - 11 1824 107
          NM_001301253.1 NP_001288182.1 11 - 12 1500 107
          NM_001301254.1 NP_001288183.1 9 - 10 1620 107
          NM_145698.4 NP_663736.2 12 - 13 1756 107
          XM_006717528.3 XP_006717591.3 13 - 14 8189 107
          XM_006717530.3 XP_006717593.1 13 - 14 1788 107
          XM_006717531.2 XP_006717594.1 13 - 14 1798 107
          XM_006717535.2 XP_006717598.1 13 - 14 1655 107
          XM_017016886.1 XP_016872375.1 12 - 13 8156 107
          XM_017016892.1 XP_016872381.1 12 - 13 1672 107
          XM_017016894.1 XP_016872383.1 11 - 12 7952 107
          XM_017016897.1 XP_016872386.1 12 - 13 1580 107
          XM_017016899.1 XP_016872388.1 11 - 12 1547 107
          XM_017016902.1 XP_016872391.1 12 - 13 1572 107
          XM_017016903.1 XP_016872392.1 12 - 13 1557 107
          XM_017016906.1 XP_016872395.1 11 - 12 1419 107
          XM_017016907.1 XP_016872396.1 11 - 12 1552 107
          XM_017016908.1 XP_016872397.1 11 - 12 1541 107
          GenBank mRNA AB082527.1 - 11 - 12 1459 107
          AF505653.1 - 11 - 12 1497 107
          AK057469.1 - 12 - 13 1623 107
          AK294510.1 - 9 - 10 1538 107
          AK295031.1 - 11 - 12 1582 107
          AK304842.1 - 10 - 11 1475 107
          AL133064.1 - 6 - 7 839 107
          BC030555.1 - 12 - 13 1660 5218685 107

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene ACBD5

          Panther Classification:

          Molecular Function -

          transfer/carrier protein

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          macroautophagy
          pexophagy
          aggrephagy
          fatty-acyl-CoA binding
          molecular_function
          lipid binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline