| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007466.hg.1 |
ENST00000429010.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007466.hg.1 |
ENST00000429010.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007466.hg.1 |
ENST00000429010.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007466.hg.1 |
ENST00000429010.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007466.hg.1 |
NR_108090.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007466.hg.1 |
NR_108090.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007466.hg.1 |
NR_108090.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007466.hg.1 |
NR_108090.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010328.hg.1 |
ENST00000507759.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010328.hg.1 |
ENST00000507759.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010328.hg.1 |
ENST00000507759.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010328.hg.1 |
ENST00000507759.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010328.hg.1 |
ENST00000507759.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010328.hg.1 |
ENST00000507759.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010328.hg.1 |
ENST00000507759.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010328.hg.1 |
ENST00000515720.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010328.hg.1 |
ENST00000515720.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010328.hg.1 |
ENST00000515720.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010328.hg.1 |
ENST00000515720.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010328.hg.1 |
ENST00000515720.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007466.hg.1 |
ENST00000429010.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007466.hg.1 |
ENST00000429010.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007466.hg.1 |
ENST00000429010.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007466.hg.1 |
ENST00000429010.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007466.hg.1 |
NR_108090.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700007466.hg.1 |
NR_108090.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700007466.hg.1 |
NR_108090.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700007466.hg.1 |
NR_108090.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700151923.hg.1 |
ENST00000429010.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700151923.hg.1 |
NR_108090.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700151923.hg.1 |
ENST00000429010.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700151923.hg.1 |
NR_108090.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700151923.hg.1 |
TC0700007466.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0400010328.hg.1 |
ENST00000507759.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010328.hg.1 |
ENST00000507759.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010328.hg.1 |
ENST00000507759.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010328.hg.1 |
ENST00000507759.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010328.hg.1 |
ENST00000507759.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010328.hg.1 |
ENST00000507759.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010328.hg.1 |
ENST00000507759.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010328.hg.1 |
ENST00000515720.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010328.hg.1 |
ENST00000515720.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010328.hg.1 |
ENST00000515720.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010328.hg.1 |
ENST00000515720.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010328.hg.1 |
ENST00000515720.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700151924.hg.1 |
ENST00000429010.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700151924.hg.1 |
NR_108090.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700151924.hg.1 |
ENST00000429010.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700151924.hg.1 |
NR_108090.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700151924.hg.1 |
TC0700007466.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0700151925.hg.1 |
ENST00000429010.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700151925.hg.1 |
NR_108090.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700151925.hg.1 |
ENST00000429010.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700151925.hg.1 |
NR_108090.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700151925.hg.1 |
TC0700007466.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0700151926.hg.1 |
ENST00000429010.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700151926.hg.1 |
NR_108090.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700151926.hg.1 |
ENST00000429010.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700151926.hg.1 |
NR_108090.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700151926.hg.1 |
TC0700007466.hg.1 |
Clariom D TC |