Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04409576_s1
          See other MMP24-AS1 GE Assays ›
          Gene Symbol:
          MMP24-AS1
          Gene Name:
          MMP24 antisense RNA 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.20: 35276333 - 35278157 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04409576_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price 434.00
          Your Price
          Online offer: 407.65
          407.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: MMP24-AS1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_102705.1 2 816 104
          NR_102706.1 2 779 104
          GenBank mRNA BU159548.1 - 1 59 104
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BG338858.1 - 1 377 Chr. 20 - 35276935 - 35277641 [-] on Build GRCh38 104
          GenBankmRNA BG469257.1 - 290 Chr. - - [-] on Build 104
          GenBankmRNA AW136076.1 - 1 Chr. 20 - 35276934 - 35277358 [+] on Build GRCh38 104
          GenBankmRNA BM765262.1 - 1 245 Chr. 20 - 35276947 - 35277510 [-] on Build GRCh38 104
          GenBankmRNA HY218680.1 - 1 Chr. 20 - 35276961 - 35277386 [+] on Build GRCh38 104
          GenBankmRNA BM826523.1 - 1 80 Chr. 20 - 35276693 - 35277345 [-] on Build GRCh38 104
          GenBankmRNA HY299780.1 - 1 Chr. 20 - 35276961 - 35277403 [+] on Build GRCh38 104
          GenBankmRNA BM806270.1 - Chr. - - [-] on Build 104
          GenBankmRNA CA866441.1 - 1 42 Chr. 20 - 35276686 - 35277304 [-] on Build GRCh38 104
          GenBankmRNA HY327436.1 - 1 Chr. 20 - 35277117 - 35277564 [+] on Build GRCh38 104
          GenBankmRNA AI242330.1 - 1 Chr. 20 - 35276946 - 35277358 [+] on Build GRCh38 105
          GenBankmRNA AA595104.1 - 1 Chr. 20 - 35276938 - 35277381 [+] on Build GRCh38 104
          GenBankmRNA AI074858.1 - 1 Chr. 20 - 35276934 - 35277351 [+] on Build GRCh38 104
          GenBankmRNA BM806312.1 - Chr. - - [-] on Build 104
          GenBankmRNA CB305660.1 - 1 Chr. 20 - 35276945 - 35277561 [+] on Build GRCh38 104
          GenBankmRNA BF026236.1 - 1 201 Chr. 20 - 35276696 - 35277467 [-] on Build GRCh38 104
          GenBankmRNA BQ887586.1 - 1 465 Chr. 20 - 35276823 - 35277730 [-] on Build GRCh38 104
          GenBankmRNA BF684267.1 - 1 375 Chr. 20 - 35276855 - 35277641 [-] on Build GRCh38 104
          GenBankmRNA CD743252.1 - 2 Chr. 20 - 35245449 - 35277524 [+] on Build GRCh38 104
          GenBankmRNA BI335118.1 - 1 51 Chr. 20 - 35276571 - 35277316 [-] on Build GRCh38 104
          GenBankmRNA BU148567.1 - 1 465 Chr. 20 - 35276833 - 35277730 [-] on Build GRCh38 104

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream metallothionein 1 pseudogene 3 MT1P3 C20orf127,
          MTL4,
          dJ614O4.6
          -58.1kb Anti-sense Chr. 20 - 35218009 - 35218185 on NCBI Build GRCh38
          Downstream eukaryotic translation initiation factor 6 EIF6 CAB,
          EIF3A,
          ITGB4BP,
          b(2)gcn,
          eIF-6,
          p27(BBP),
          p27BBP
          974.0 Sense Chr. 20 - 35278906 - 35284816 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC2000010005.hg.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC2000010005.hg.1 ENST00000424358.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000010005.hg.1 ENST00000424358.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000010005.hg.1 ENST00000424358.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000010005.hg.1 ENST00000424358.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000010005.hg.1 ENST00000424358.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000010005.hg.1 ENST00000435366.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000010005.hg.1 ENST00000435366.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000010005.hg.1 ENST00000435366.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000010005.hg.1 ENST00000435366.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000010005.hg.1 ENST00000435366.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000010005.hg.1 ENST00000435366.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000010005.hg.1 ENST00000456790.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000010005.hg.1 ENST00000456790.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000010005.hg.1 ENST00000456790.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000010005.hg.1 NR_102705.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000010005.hg.1 NR_102705.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000010005.hg.1 NR_102705.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000010005.hg.1 NR_102705.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000010005.hg.1 NR_102705.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000010005.hg.1 NR_102705.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000010005.hg.1 NR_102705.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000010005.hg.1 NR_102706.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000010005.hg.1 NR_102706.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000010005.hg.1 NR_102706.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000010005.hg.1 NR_102706.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000010005.hg.1 NR_102706.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000010005.hg.1 NR_102706.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000010005.hg.1 NR_102706.1 Refseq
          Affymetrix Human Clariom D Assay PSR2000171454.hg.1 ENST00000424358.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2000171454.hg.1 ENST00000456790.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2000171454.hg.1 NR_102705.1 Refseq
          Affymetrix Human Clariom D Assay PSR2000171454.hg.1 NR_102706.1 Refseq
          Affymetrix Human Clariom D Assay PSR2000171454.hg.1 TC2000010005.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2000171454.hg.1 ENST00000424358.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2000171454.hg.1 ENST00000456790.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2000171454.hg.1 NR_102705.1 Refseq
          Affymetrix Human Clariom D Assay PSR2000171454.hg.1 NR_102706.1 Refseq
          Affymetrix Human Clariom D Assay PSR2000171455.hg.1 ENST00000424358.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2000171455.hg.1 ENST00000435366.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2000171455.hg.1 ENST00000435366.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2000171455.hg.1 ENST00000456790.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2000171455.hg.1 NR_102705.1 Refseq
          Affymetrix Human Clariom D Assay PSR2000171455.hg.1 NR_102706.1 Refseq
          Affymetrix Human Clariom D Assay PSR2000171455.hg.1 TC2000010005.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2000171455.hg.1 ENST00000424358.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2000171455.hg.1 ENST00000435366.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2000171455.hg.1 ENST00000435366.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2000171455.hg.1 ENST00000456790.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2000171455.hg.1 NR_102705.1 Refseq
          Affymetrix Human Clariom D Assay PSR2000171455.hg.1 NR_102706.1 Refseq
          Affymetrix Human Clariom D Assay TC2000010005.hg.1 ENST00000424358.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000010005.hg.1 ENST00000424358.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000010005.hg.1 ENST00000424358.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000010005.hg.1 ENST00000424358.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000010005.hg.1 ENST00000424358.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000010005.hg.1 ENST00000435366.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000010005.hg.1 ENST00000435366.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000010005.hg.1 ENST00000435366.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000010005.hg.1 ENST00000435366.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000010005.hg.1 ENST00000435366.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000010005.hg.1 ENST00000435366.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000010005.hg.1 ENST00000456790.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000010005.hg.1 ENST00000456790.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000010005.hg.1 ENST00000456790.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000010005.hg.1 NR_102705.1 Refseq
          Affymetrix Human Clariom D Assay TC2000010005.hg.1 NR_102705.1 Refseq
          Affymetrix Human Clariom D Assay TC2000010005.hg.1 NR_102705.1 Refseq
          Affymetrix Human Clariom D Assay TC2000010005.hg.1 NR_102705.1 Refseq
          Affymetrix Human Clariom D Assay TC2000010005.hg.1 NR_102705.1 Refseq
          Affymetrix Human Clariom D Assay TC2000010005.hg.1 NR_102705.1 Refseq
          Affymetrix Human Clariom D Assay TC2000010005.hg.1 NR_102705.1 Refseq
          Affymetrix Human Clariom D Assay TC2000010005.hg.1 NR_102706.1 Refseq
          Affymetrix Human Clariom D Assay TC2000010005.hg.1 NR_102706.1 Refseq
          Affymetrix Human Clariom D Assay TC2000010005.hg.1 NR_102706.1 Refseq
          Affymetrix Human Clariom D Assay TC2000010005.hg.1 NR_102706.1 Refseq
          Affymetrix Human Clariom D Assay TC2000010005.hg.1 NR_102706.1 Refseq
          Affymetrix Human Clariom D Assay TC2000010005.hg.1 NR_102706.1 Refseq
          Affymetrix Human Clariom D Assay TC2000010005.hg.1 NR_102706.1 Refseq
          Affymetrix Human Clariom D Assay PSR2000171458.hg.1 ENST00000424358.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2000171458.hg.1 ENST00000456790.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2000171458.hg.1 NR_102705.1 Refseq
          Affymetrix Human Clariom D Assay PSR2000171458.hg.1 NR_102706.1 Refseq
          Affymetrix Human Clariom D Assay PSR2000171458.hg.1 TC2000010005.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2000171458.hg.1 ENST00000424358.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2000171458.hg.1 ENST00000456790.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2000171458.hg.1 NR_102705.1 Refseq
          Affymetrix Human Clariom D Assay PSR2000171458.hg.1 NR_102706.1 Refseq
          Affymetrix Human Clariom D Assay PSR2000171453.hg.1 ENST00000424358.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2000171453.hg.1 NR_102705.1 Refseq
          Affymetrix Human Clariom D Assay PSR2000171453.hg.1 NR_102706.1 Refseq
          Affymetrix Human Clariom D Assay PSR2000171453.hg.1 TC2000010005.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2000171453.hg.1 ENST00000424358.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2000171453.hg.1 NR_102705.1 Refseq
          Affymetrix Human Clariom D Assay PSR2000171453.hg.1 NR_102706.1 Refseq
          Affymetrix Human Clariom D Assay PSR2000171459.hg.1 NR_102705.1 Refseq
          Affymetrix Human Clariom D Assay PSR2000171459.hg.1 NR_102706.1 Refseq
          Affymetrix Human Clariom D Assay PSR2000171459.hg.1 TC2000010005.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2000171459.hg.1 NR_102705.1 Refseq
          Affymetrix Human Clariom D Assay PSR2000171459.hg.1 NR_102706.1 Refseq
          Affymetrix Human Clariom D Assay PSR2000171451.hg.1 ENST00000435366.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2000171451.hg.1 ENST00000435366.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2000171451.hg.1 NR_102705.1 Refseq
          Affymetrix Human Clariom D Assay PSR2000171451.hg.1 NR_102706.1 Refseq
          Affymetrix Human Clariom D Assay PSR2000171451.hg.1 TC2000010005.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2000171451.hg.1 ENST00000435366.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2000171451.hg.1 ENST00000435366.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2000171451.hg.1 NR_102705.1 Refseq
          Affymetrix Human Clariom D Assay PSR2000171451.hg.1 NR_102706.1 Refseq
          Affymetrix Human Clariom D Assay PSR2000171452.hg.1 ENST00000424358.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2000171452.hg.1 ENST00000435366.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2000171452.hg.1 ENST00000435366.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2000171452.hg.1 NR_102705.1 Refseq
          Affymetrix Human Clariom D Assay PSR2000171452.hg.1 NR_102706.1 Refseq
          Affymetrix Human Clariom D Assay PSR2000171452.hg.1 TC2000010005.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2000171452.hg.1 ENST00000424358.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2000171452.hg.1 ENST00000435366.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2000171452.hg.1 ENST00000435366.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2000171452.hg.1 NR_102705.1 Refseq
          Affymetrix Human Clariom D Assay PSR2000171452.hg.1 NR_102706.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline