Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04931354_mH
          See other TNFRSF10D GE Assays ›
          Gene Symbol:
          TNFRSF10D
          Gene Name:
          TNF receptor superfamily member 10d
          Gene Aliases:
          CD264, DCR2, TRAIL-R4, TRAILR4, TRUNDD
          Chromosome Location:
          Chr.8: 23135588 - 23164030 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04931354_mH
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: TNFRSF10D
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_003840.4 NP_003831.2 4 - 5 580 108
          GenBank mRNA AF021232.1 - 4 - 5 563 108
          AF021233.1 - 4 - 5 564 108
          AF023849.1 - 4 - 5 485 108
          AF029761.1 - 4 - 5 567 108
          AK313528.1 - 4 - 5 577 108
          AY358285.1 - 4 - 5 568 108
          BC052270.1 - 4 - 5 568 6572696 108

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0800009872.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC08001051.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene TNFRSF10D

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          inflammatory response
          immune response
          signal transduction
          multicellular organism development
          extrinsic apoptotic signaling pathway via death domain receptors
          response to lipopolysaccharide
          tumor necrosis factor-mediated signaling pathway
          regulation of cell proliferation
          regulation of apoptotic process
          negative regulation of apoptotic process
          transmembrane signaling receptor activity
          tumor necrosis factor-activated receptor activity
          TRAIL binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0800009872.hg.1 ENST00000312584.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009872.hg.1 ENST00000312584.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009872.hg.1 ENST00000312584.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009872.hg.1 ENST00000312584.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009872.hg.1 ENST00000312584.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009872.hg.1 ENST00000312584.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009872.hg.1 ENST00000312584.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009872.hg.1 ENST00000312584.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009872.hg.1 ENST00000312584.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009872.hg.1 ENST00000312584.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009872.hg.1 ENST00000312584.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009872.hg.1 ENST00000312584.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009872.hg.1 NM_003840.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800009872.hg.1 NM_003840.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800009872.hg.1 NM_003840.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800009872.hg.1 NM_003840.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800009872.hg.1 NM_003840.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800009872.hg.1 NM_003840.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800009872.hg.1 NM_003840.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800009872.hg.1 NM_003840.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800009872.hg.1 NM_003840.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800009872.hg.1 NM_003840.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800009872.hg.1 NM_003840.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800009872.hg.1 NM_003840.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR08013373.hg.1 ENST00000312584.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08013373.hg.1 NM_003840.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR08013373.hg.1 TC08001051.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0800167026.hg.1 ENST00000312584.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0800167026.hg.1 NM_003840.4 Refseq
          Affymetrix Human Clariom D Assay PSR0800167026.hg.1 ENST00000312584.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0800167026.hg.1 NM_003840.4 Refseq
          Affymetrix Human Clariom D Assay PSR0800167026.hg.1 TC0800009872.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0800009872.hg.1 ENST00000312584.3 Ensembl
          Affymetrix Human Clariom D Assay TC0800009872.hg.1 ENST00000312584.3 Ensembl
          Affymetrix Human Clariom D Assay TC0800009872.hg.1 ENST00000312584.3 Ensembl
          Affymetrix Human Clariom D Assay TC0800009872.hg.1 ENST00000312584.3 Ensembl
          Affymetrix Human Clariom D Assay TC0800009872.hg.1 ENST00000312584.3 Ensembl
          Affymetrix Human Clariom D Assay TC0800009872.hg.1 ENST00000312584.3 Ensembl
          Affymetrix Human Clariom D Assay TC0800009872.hg.1 ENST00000312584.3 Ensembl
          Affymetrix Human Clariom D Assay TC0800009872.hg.1 ENST00000312584.3 Ensembl
          Affymetrix Human Clariom D Assay TC0800009872.hg.1 ENST00000312584.3 Ensembl
          Affymetrix Human Clariom D Assay TC0800009872.hg.1 ENST00000312584.3 Ensembl
          Affymetrix Human Clariom D Assay TC0800009872.hg.1 ENST00000312584.3 Ensembl
          Affymetrix Human Clariom D Assay TC0800009872.hg.1 ENST00000312584.3 Ensembl
          Affymetrix Human Clariom D Assay TC0800009872.hg.1 NM_003840.4 Refseq
          Affymetrix Human Clariom D Assay TC0800009872.hg.1 NM_003840.4 Refseq
          Affymetrix Human Clariom D Assay TC0800009872.hg.1 NM_003840.4 Refseq
          Affymetrix Human Clariom D Assay TC0800009872.hg.1 NM_003840.4 Refseq
          Affymetrix Human Clariom D Assay TC0800009872.hg.1 NM_003840.4 Refseq
          Affymetrix Human Clariom D Assay TC0800009872.hg.1 NM_003840.4 Refseq
          Affymetrix Human Clariom D Assay TC0800009872.hg.1 NM_003840.4 Refseq
          Affymetrix Human Clariom D Assay TC0800009872.hg.1 NM_003840.4 Refseq
          Affymetrix Human Clariom D Assay TC0800009872.hg.1 NM_003840.4 Refseq
          Affymetrix Human Clariom D Assay TC0800009872.hg.1 NM_003840.4 Refseq
          Affymetrix Human Clariom D Assay TC0800009872.hg.1 NM_003840.4 Refseq
          Affymetrix Human Clariom D Assay TC0800009872.hg.1 NM_003840.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-454p7:80/100.66.72.101:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline