Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04963596_m1
          See other SALL1 GE Assays ›
          Gene Symbol:
          SALL1
          Gene Name:
          spalt like transcription factor 1
          Gene Aliases:
          HEL-S-89, HSAL1, Sal-1, TBS, ZNF794
          Chromosome Location:
          Chr.16: 51135975 - 51152386 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04963596_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SALL1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001127892.1 NP_001121364.1 1 - 2 219 76
          GenBank mRNA AK307835.1 - 1 - 2 219 76

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the NM_001127892.1 transcripts.

          Set Membership:

          Amplicon length between 71 and 85 Homo sapiens Probe spans exons

          Panther Classification:

          Molecular Function -

          C2H2 zinc finger transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          ureteric bud development
          branching involved in ureteric bud morphogenesis
          kidney development
          ventricular septum development
          mesenchymal to epithelial transition involved in metanephros morphogenesis
          transcription, DNA-templated
          heart development
          gonad development
          histone deacetylation
          olfactory nerve development
          olfactory bulb interneuron differentiation
          pituitary gland development
          neurogenesis
          positive regulation of Wnt signaling pathway
          adrenal gland development
          inductive cell-cell signaling
          somatic stem cell population maintenance
          outer ear morphogenesis
          embryonic digit morphogenesis
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          embryonic digestive tract development
          limb development
          olfactory bulb mitral cell layer development
          kidney epithelium development
          ureteric bud invasion
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          histone deacetylase activity
          protein binding
          beta-catenin binding
          sequence-specific DNA binding
          transcription regulatory region DNA binding
          metal ion binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1600010291.hg.1 ENST00000251020.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010291.hg.1 ENST00000251020.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010291.hg.1 ENST00000251020.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010291.hg.1 ENST00000251020.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010291.hg.1 ENST00000251020.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010291.hg.1 ENST00000251020.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010291.hg.1 ENST00000251020.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010291.hg.1 ENST00000251020.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010291.hg.1 ENST00000440970.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010291.hg.1 ENST00000440970.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010291.hg.1 ENST00000440970.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010291.hg.1 ENST00000440970.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010291.hg.1 ENST00000440970.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010291.hg.1 ENST00000440970.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010291.hg.1 ENST00000440970.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010291.hg.1 ENST00000562674.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010291.hg.1 ENST00000562674.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010291.hg.1 ENST00000562674.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010291.hg.1 ENST00000562674.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010291.hg.1 ENST00000566102.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010291.hg.1 ENST00000566102.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010291.hg.1 ENST00000566102.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010291.hg.1 ENST00000566102.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010291.hg.1 ENST00000570206.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010291.hg.1 ENST00000570206.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010291.hg.1 ENST00000570206.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010291.hg.1 ENST00000570206.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010291.hg.1 NM_001127892.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600010291.hg.1 NM_001127892.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600010291.hg.1 NM_001127892.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600010291.hg.1 NM_001127892.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600010291.hg.1 NM_001127892.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600010291.hg.1 NM_001127892.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600010291.hg.1 NM_001127892.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600010291.hg.1 NM_002968.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600010291.hg.1 NM_002968.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600010291.hg.1 NM_002968.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600010291.hg.1 NM_002968.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600010291.hg.1 NM_002968.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600010291.hg.1 NM_002968.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600010291.hg.1 NM_002968.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600010291.hg.1 NM_002968.2 Refseq
          Affymetrix Human Clariom D Assay TC1600010291.hg.1 ENST00000251020.8 Ensembl
          Affymetrix Human Clariom D Assay TC1600010291.hg.1 ENST00000251020.8 Ensembl
          Affymetrix Human Clariom D Assay TC1600010291.hg.1 ENST00000251020.8 Ensembl
          Affymetrix Human Clariom D Assay TC1600010291.hg.1 ENST00000251020.8 Ensembl
          Affymetrix Human Clariom D Assay TC1600010291.hg.1 ENST00000251020.8 Ensembl
          Affymetrix Human Clariom D Assay TC1600010291.hg.1 ENST00000251020.8 Ensembl
          Affymetrix Human Clariom D Assay TC1600010291.hg.1 ENST00000251020.8 Ensembl
          Affymetrix Human Clariom D Assay TC1600010291.hg.1 ENST00000251020.8 Ensembl
          Affymetrix Human Clariom D Assay TC1600010291.hg.1 ENST00000440970.5 Ensembl
          Affymetrix Human Clariom D Assay TC1600010291.hg.1 ENST00000440970.5 Ensembl
          Affymetrix Human Clariom D Assay TC1600010291.hg.1 ENST00000440970.5 Ensembl
          Affymetrix Human Clariom D Assay TC1600010291.hg.1 ENST00000440970.5 Ensembl
          Affymetrix Human Clariom D Assay TC1600010291.hg.1 ENST00000440970.5 Ensembl
          Affymetrix Human Clariom D Assay TC1600010291.hg.1 ENST00000440970.5 Ensembl
          Affymetrix Human Clariom D Assay TC1600010291.hg.1 ENST00000440970.5 Ensembl
          Affymetrix Human Clariom D Assay TC1600010291.hg.1 ENST00000562674.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010291.hg.1 ENST00000562674.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010291.hg.1 ENST00000562674.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010291.hg.1 ENST00000562674.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010291.hg.1 ENST00000566102.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010291.hg.1 ENST00000566102.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010291.hg.1 ENST00000566102.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010291.hg.1 ENST00000566102.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010291.hg.1 ENST00000570206.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010291.hg.1 ENST00000570206.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010291.hg.1 ENST00000570206.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010291.hg.1 ENST00000570206.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010291.hg.1 NM_001127892.1 Refseq
          Affymetrix Human Clariom D Assay TC1600010291.hg.1 NM_001127892.1 Refseq
          Affymetrix Human Clariom D Assay TC1600010291.hg.1 NM_001127892.1 Refseq
          Affymetrix Human Clariom D Assay TC1600010291.hg.1 NM_001127892.1 Refseq
          Affymetrix Human Clariom D Assay TC1600010291.hg.1 NM_001127892.1 Refseq
          Affymetrix Human Clariom D Assay TC1600010291.hg.1 NM_001127892.1 Refseq
          Affymetrix Human Clariom D Assay TC1600010291.hg.1 NM_001127892.1 Refseq
          Affymetrix Human Clariom D Assay TC1600010291.hg.1 NM_002968.2 Refseq
          Affymetrix Human Clariom D Assay TC1600010291.hg.1 NM_002968.2 Refseq
          Affymetrix Human Clariom D Assay TC1600010291.hg.1 NM_002968.2 Refseq
          Affymetrix Human Clariom D Assay TC1600010291.hg.1 NM_002968.2 Refseq
          Affymetrix Human Clariom D Assay TC1600010291.hg.1 NM_002968.2 Refseq
          Affymetrix Human Clariom D Assay TC1600010291.hg.1 NM_002968.2 Refseq
          Affymetrix Human Clariom D Assay TC1600010291.hg.1 NM_002968.2 Refseq
          Affymetrix Human Clariom D Assay TC1600010291.hg.1 NM_002968.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline