Search
Search

PMF1-BGLAP| Gene Symbol: PMF1-BGLAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199661.1 | NP_001186590.1 | 4 - 5 | 536 | 102 | |
| NM_001199662.1 | NP_001186591.1 | 4 - 5 | 597 | 102 | ||
| NM_001199664.1 | NP_001186593.1 | 2 - 3 | 390 | 102 | ||
| GenBank mRNA | CD049192.1 | - | 4 - 5 | 605 | 102 | |
| CD388852.1 | - | 4 - 5 | 536 | 102 | ||
| FY213424.1 | - | 2 - 3 | 303 | 102 | ||
Set Membership: |
|