Search
Search

CALML6| Gene Symbol: CALML6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015443822.1 | XP_015299308.1 | 3 - 4 | 630 | 103 | |
| XM_015443824.1 | XP_015299310.1 | 3 - 4 | 615 | 103 | ||
| XM_015443825.1 | XP_015299311.1 | 4 - 5 | 2567 | 103 | ||
| XM_015443827.1 | XP_015299313.1 | 4 - 5 | 2552 | 103 | ||
Set Membership: |
|